Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA049580

Sigma-Aldrich

Anti-CDX2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

CDX3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

PGPLRSVPEPLSPVSSLQASVPGSVPGVLGPTGGVLNPTVT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDX2(1045)

General description

Caudal type homeobox 2 (CDX2) is a member of caudal-related homeobox transcription factor gene family. Cdx2 is homologous to Drosophila developmental gene caudal. The Cdx2 gene is located on the human chromosome on 13q12.2. It is prominently produced in the intestine.

Immunogen

caudal type homeobox 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CDX2 antibody recognises CDX2 protein.

Biochem/physiol Actions

Caudal type homeobox 2 (CDX2) plays a key role during early developmental stages by providing positional information for gastrointestinal tract, especially the midgut region. Cdx2 promotes proliferation and differentiation of intestinal epithelial cells. Cdx2 has tumour suppression function in colon cancer cells. Cdx2 expression level is found to decrease as the cancer advances from dysplasia to advanced gastric cancer. Abnormal amplification and upregulation of Cdx2 leads to colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST91202

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Cdx2 homeobox gene has a tumour suppressor function in the distal colon in addition to a homeotic role during gut development
Bonhomme C, et al.
Gut, 52(10), 1465-1471 (2003)
Cloning and chromosome assignment of the human CDX2 gene
Drummond F, et al.
Annals of Human Genetics, 61(5), 393-400 (1997)
Vasty Osei Amponsa et al.
American journal of clinical and experimental urology, 7(3), 139-152 (2019-07-19)
Creation of genetically engineered mouse models of bladder cancer often involves the use of several background strains in conjunction with the carcinogen N-butyl-N-(4-hydroxybutyl) nitrosamine (BBN). However, carcinogen susceptibility in commonly used strains, as well as phenotypic differences is not well
CDX2 is an amplified lineage-survival oncogene in colorectal cancer
Salari K, et al.
Proceedings of the National Academy of Sciences of the USA, 61(5), 201206004-201206004 (2012)
CDX2 expression in the intestinal-type gastric epithelial neoplasia: frequency and significance
Srivastava A, et al.
Modern Pathology, 23(1), 54-54 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service