Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA042294

Sigma-Aldrich

Anti-CENPE antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Centromere protein e, 312kda, Anti-Kif10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

EKIENLTRMLVTSSSLTLQQELKAKRKRRVTWCLGKINKMKNSNYADQFNIPTNITTKTHKLSINLLREIDESVCSESDVFSNTLDTLSEIEWNPATKLLNQENIESELNSLRADYDN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CENPE(1062)

General description

The gene CENPE (centromere protein E) is mapped to human chromosome 4q24. The encoded protein belongs to the kinesin-7 family of proteins. The CENPE protein is present with spindle microtubules. The protein has a catalytic motor domain, a discontinuous a-helical coiled-coil domain and an ATP-independent microtubule-interacting domain.

Immunogen

centromere protein E, 312kDa recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CENPE (centromere protein E) is a plus-end-directed mitotic kinesin which is required for a stable connection between kinetochores and spindle microtubules. It plays an important role in the movement of duplicated chromosomal pairs. It also prevents aneuploidy. Absence of CENPE results in misalignment of chromosomes at the spindle pole. Mutation in this gene is associated with microcephalic primordial dwarfism. Absence of CENPE in mice causes embryonic lethality.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71072

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosome congression is promoted by CENP-Q- and CENP-E-dependent pathways.
Bancroft J, et al.
Journal of Cell Science, 128, 171-184 (2015)
Common variants identified in genome-wide association studies of testicular germ cell tumour: an update, biological insights and clinical application.
Litchfield K, et al.
Andrology, 3, 34-46 (2015)
Mitosis. Microtubule detyrosination guides chromosomes during mitosis.
Barisic M, et al.
Science, 348, 799-803 (2015)
Mutations in CENPE define a novel kinetochore-centromeric mechanism for microcephalic primordial dwarfism.
Mirzaa GM, et al.
Human Genetics, 133, 1023-1039 (2014)
Tiaojiang Xiao et al.
Cell reports, 12(10), 1704-1714 (2015-09-01)
The role of CTCF in stabilizing long-range interactions between chromatin sites essential for maintaining nuclear architecture is well established. Most of these interactions involve recruitment of the cohesin complex to chromatin via CTCF. We find that CTCF also interacts with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service