Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA026803

Sigma-Aldrich

Anti-RNF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BAP-1, Anti-BAP1, Anti-DING, Anti-HIPI3, Anti-RING1B, Anti-RING2, Anti-ring finger protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ALISKIYPSRDEYEAHQERVLARINKHNNQQALSHSIEEGLKIQAMNRLQRGKKQQIENGSGAEDNGDSSHCSNASTHSNQEAGPSNKRTKTSDDSGLELDNNNAAMAIDPVMDGASEIELV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF2(6045)

General description

RING finger protein 2 (RNF2), also known as RNF1b, consists of 336 amino acids and it belongs to PcG (polycomb-group) protein family. This protein is encoded by the gene mapped on human chromosome 1q25.3. The encoded protein is characterized with a unique C3HC4 RING finger motif.

Immunogen

ring finger protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RING finger protein 2 (RNF2), in association with prohibitin, regulates the activity of E2F transcription factor 1(E2F1) by the direct, prohibitin-mediated pathway and the indirect, p16-mediated pathway. Rnf2 might act as a vital assembly factor in PRC (Polycomb repressive complex), which is required for mammalian development. Decrease in the expression of Rnf2 or ring1b causes gastrulation arrest and cell cycle inhibition. Overexpression of RNF2 results in pathogenesis of urothelial carcinoma of the bladder (UCB). RNF2 interacts with E2 ubiquitin-conjugating enzymes and display E3 ubiquitin ligase activity. RNF2 plays a crucial role in cancer cell survival and proliferation, therefore it can serve as a potential therapeutic target for cancer treatment and prevention.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71062

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of RNF2 Is an Independent Predictor of Outcome in Patients with Urothelial Carcinoma of the Bladder Undergoing Radical Cystectomy.
Li XD
Scientific Reports, 6:20894 (2016)
Knockdown of RNF2 induces apoptosis by regulating MDM2 and p53 stability.
Wen W
Oncogene, 33(4), 421-428 (2014)
Prohibitin interacts with RNF2 and regulates E2F1 function via dual pathways.
Choi D
Oncogene, 27(12), 1716-1725 (2008)
RNF2 interacts with the linker region of the human P-glycoprotein.
Rao PS
International Journal of Oncology, 29(6), 1413-1419 (2006)
Łukasz Kuźbicki et al.
Oncotarget, 9(25), 17656-17663 (2018-05-01)
Recent studies involving melanoma cell lines suggest that enhanced expression of epigenetic regulator RNF2 supports proliferation and promotes metastasis. However, it is not clear to what extent those data apply to disease progression and prognosis for melanoma patients. Therefore the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service