Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA025226

Sigma-Aldrich

Anti-GGH antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Conjugase, Anti-GH, Anti-Gamma-Glu-X carboxypeptidase, Anti-Gamma-glutamyl hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FDDGDYFPVWGTCLGFEELSLLISGECLLTATDTVDVAMPLNFTGGQLHSRMFQNFPTELLLSLAVEPLTANFHKWSLSVKNFTMNEKLKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGH(8836)

General description

The gene GGH (γ-glutamyl hydrolase) is mapped to human chromosome 8q12.1. The protein is mainly present in the lysosomes.

Immunogen

Gamma-glutamyl hydrolase Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-GGH antibody produced in rabbit has been used for immunohistochemistry and western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

GGH (γ-glutamyl hydrolase) is involved in the removal of γ-linked polyglutamates, thereby hydrolysing (anti)folylpoly-γ-glutamates. The enzyme controls the intracellular folate pools and folate metabolism. During the treatment of acute lymphoblastic leukemia (ALL), mutation in the GGH gene is linked with efficacy and side effects of methotrexate (MTX). It is responsible for changing a long-chain methotrexate polyglutamates (MTX-PGs) into a short-chain MTX-PGs and finally to MTX. Mutation in this gene at -401C>T enhances the response of cisplatin-based chemoradiotherapy in cervical carcinoma. The levels of GGH are higher in invasive breast cancer tumors and are associated with the development and progression of the cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74642

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel Melling et al.
International journal of molecular sciences, 18(2) (2017-02-02)
γ-glutamyl-hydrolase (GGH) is a ubiquitously-expressed enzyme that regulates intracellular folate metabolism for cell proliferation, DNA synthesis, and repair. Employing GGH immunohistochemistry on a tissue microarray with 12,427 prostate cancers, we found that GGH expression was negative to low in normal
Proteomic analysis of human substantia nigra identifies novel candidates involved in Parkinson's disease pathogenesis.
Licker V, et al.
Proteomics, 14, 784-794 (2014)
Identification of carboxypeptidase E and gamma-glutamyl hydrolase as biomarkers for pulmonary neuroendocrine tumors by cDNA microarray.
He P, et al.
Human Pathology, 35, 1196-1209 (2004)
Virginie Licker et al.
Proteomics, 14(6), 784-794 (2014-01-23)
Parkinson's disease (PD) pathology spreads throughout the brain following a region-specific process predominantly affecting the substantia nigra (SN) pars compacta. SN exhibits a progressive loss of dopaminergic neurons responsible for the major cardinal motor symptoms, along with the occurrence of
Alessandro Bertolo et al.
Scientific reports, 9(1), 2074-2074 (2019-02-16)
Mesenchymal stromal cells (MSC) are used in cell therapies, however cellular senescence increases heterogeneity of cell populations and leads to uncertainty in therapies' outcomes. The determination of cellular senescence is time consuming and logistically intensive. Here, we propose the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service