Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA021257

Sigma-Aldrich

Anti-BAIAP2L1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-BAI1-associated protein 2-like protein 1, Anti-Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1, Anti-Insulin receptor tyrosine kinase substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LLSFAQGDVITLLIPEEKDGWLYGEHDVSKARGWFPSSYTKLLEENETEAVTVPTPSPTPVRSISTVNLSEN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

BAIAP2L1 (brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1) belongs to the IRSp53 (insulin receptor substrate protein of 53kDa) family. BAIAP2L1 is widely distributed in human tissues. The protein mainly localizes in the nucleus. BAIAP2L1 is commonly referred to as IRTKS (insulin receptor tyrosine kinase substrate).

Immunogen

Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BAIAP2L1 (brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1) is linked with actin remodeling and membrane protrusion. It is up-regulated in hepatocellular carcinoma (HCC) and is associated with tumor size. In HCC cells, it promotes EGFR (epidermal growth factor receptor)-ERK (extracellular signal-regulated kinase) signaling pathway. BAIAP2L1 also inhibits p53-mediated apoptosis and transactivation activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74834

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Ping Wang et al.
Cancer letters, 337(1), 96-106 (2013-05-23)
Insulin receptor tyrosine kinase substrate (IRTKS) is closely associated with actin remodelling and membrane protrusion, but its role in the pathogenesis of malignant tumours, including hepatocellular carcinoma (HCC), is still unknown. In this study, we showed that IRTKS was frequently
Meagan M Postema et al.
Current biology : CB, 28(18), 2876-2888 (2018-09-11)
Transporting epithelial cells like those that line the gut build large arrays of actin-supported protrusions called microvilli, which extend from the apical surface into luminal spaces to increase functional surface area. Although critical for maintaining physiological homeostasis, mechanisms controlling the
Ke-Sheng Wang et al.
PloS one, 6(8), e23571-e23571 (2011-09-03)
The tumor suppressor p53 controls multiple cellular functions including DNA repair, cell cycle arrest and apoptosis. MDM2-mediated p53 ubiquitination affects both degradation and cytoplasmic localization of p53. Several cofactors are known to modulate MDM2-mediated p53 ubiquitination and proteasomal degradation. Here
Thomas H Millard et al.
Journal of cell science, 120(Pt 9), 1663-1672 (2007-04-14)
IRSp53 is a scaffold protein that contains an IRSp53/MIM homology domain (IMD) that bundles actin filaments and interacts with the small GTPase Rac. IRSp53 also binds to the small GTPase Cdc42 and to Scar/WAVE and Mena/VASP proteins to regulate the
Didier Vingadassalom et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(16), 6754-6759 (2009-04-16)
Enterohemorrhagic Escherichia coli O157:H7 translocates 2 effectors to trigger localized actin assembly in mammalian cells, resulting in filamentous actin "pedestals." One effector, the translocated intimin receptor (Tir), is localized in the plasma membrane and clustered upon binding the bacterial outer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service