Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA020300

Sigma-Aldrich

Anti-COG5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-13S Golgi transport complex 90 kDa subunit, Anti-COG complex subunit 5, Anti-Component of oligomeric Golgi complex 5, Anti-Conserved oligomeric Golgi complex subunit 5, Anti-GTC-90, Anti-Golgi transport complex 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SFWTNMEKLMDHIYAVCGQVQHLQKVLAKKRDPVSHICFIEEIVKDGQPEIFYTFWNSVTQALSSQFHMATNSSMFLKQAFEGEYPKLLRLYNDLWKRLQQYSQHIQGNFNASGTTDLYVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG5(10466)

Looking for similar products? Visit Product Comparison Guide

General description

Component of oligomeric golgi complex 5 (COG5) is a 90kDa protein which is a part of the conserved oligomeric golgi (COG) complex. The gene encoding it is localized on human chromosome 7q31.

Immunogen

Conserved oligomeric Golgi complex subunit 5 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)

Biochem/physiol Actions

Mutations in the gene encoding component of oligomeric golgi complex 5 (COG5) have been shown to be associated with congenital disorders of glycosylation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75057

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricie Paesold-Burda et al.
Human molecular genetics, 18(22), 4350-4356 (2009-08-20)
The conserved oligomeric Golgi (COG) complex is a tethering factor composed of eight subunits that is involved in the retrograde transport of intra-Golgi components. Deficient biosynthesis of COG subunits leads to alterations of protein trafficking along the secretory pathway and
D M Walter et al.
The Journal of biological chemistry, 273(45), 29565-29576 (1998-10-29)
Intracellular protein traffic involves a tightly regulated series of events in which a membrane-bounded vesicles bud from one compartment and are specifically targeted to the next compartment, where they dock and fuse. A cell-free system that reconstitutes vesicle trafficking between
Cortnie Hartwig et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(2), 215-233 (2020-11-20)
Rare genetic diseases preponderantly affect the nervous system causing neurodegeneration to neurodevelopmental disorders. This is the case for both Menkes and Wilson disease, arising from mutations in ATP7A and ATP7B, respectively. The ATP7A and ATP7B proteins localize to the Golgi
Avanti Gokhale et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(11), 3697-3711 (2012-03-17)
The Biogenesis of Lysosome-Related Organelles Complex 1 (BLOC-1) is a protein complex containing the schizophrenia susceptibility factor dysbindin, which is encoded by the gene DTNBP1. However, mechanisms engaged by dysbindin defining schizophrenia susceptibility pathways have not been quantitatively elucidated. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service