Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA020027

Sigma-Aldrich

Anti-RAPH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lamellipodin, Anti-PREL-2, Anti-Proline-rich EVH1 ligand 2, Anti-Protein RMO1, Anti-RAPH1, Anti-Ras-associated and pleckstrin homology domains-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

NPQPQQWSKMSVKKAPPPTRPKRNDSTRLTQAEISEQPTMATVVPQVPTSPKSSLSVQPGFLADLNRTLQRKSITRHGSLSSRMSRAEPTATMDDMALPPPPPELLSDQQKAGYGGSHISGYATLRRGPPPAPPKRDQNTKLSR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAPH1(65059)

General description

RAPH1 (Ras association (RalGDS/AF-6) and pleckstrin homology domains 1) is a RA-PH tandem domain containing protein encoding an adaptor protein termed as lamellipodin protein (Lpd). It is located on the chromosome location 2q33. It also consists of an N-terminal coiled-coil region (cc-RA-PH).

Immunogen

Ras-associated and pleckstrin homology domains-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAPH1 (Ras association (RalGDS/AF-6) and pleckstrin homology domains 1) is involved in the regulation of actin cytoskeleton assembly. It directly binds to the Ena/VASPs proteins to guide the cells in movement and lamelipodial protrusion. It modulates actin polymerization by binding to different phosphoinositol through Ras GTPase signaling pathway. RAPH1 also possesses tumorigenic characteristics. It controls the cell motility of breast tumor cells. Deleted expression of RAPH1 has been reported in various types of cancer cells such as breast carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74658

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawrence M Schopfer et al.
Molecules (Basel, Switzerland), 22(7) (2017-07-01)
Humans with the C5 genetic variant of butyrylcholinesterase (BChE) have 30-200% higher plasma BChE activity, low body weight, and shorter duration of action of the muscle relaxant succinylcholine. The C5 variant has an extra, slow-moving band of BChE activity on
Meire S Batistela et al.
Molecular biology reports, 40(12), 6613-6617 (2013-09-24)
Lamellipodin protein (Lpd), encoded by the RAPH1 gene, modulates the assembly of actin cytoskeleton through its binding to the Ena/VASPs proteins, and acts in cellular motility and lamelipodial protrusion. The region where RAPH1 gene is located (2q33) is deleted in
Yu-Chung Chang et al.
Protein & cell, 4(3), 211-219 (2013-03-14)
The adapter protein Lamellipodin (Lpd) plays an important role in cell migration. In particular, Lpd mediates lamellipodia formation by regulating actin dynamics via interacting with Ena/VASP proteins. Its RA-PH tandem domain configuration suggests that like its paralog RIAM, Lpd may
Yosuke Senju et al.
The Journal of biological chemistry, 299(5), 104571-104571 (2023-03-06)
Metastasis-suppressor 1 (MTSS1) is a membrane-interacting scaffolding protein that regulates the integrity of epithelial cell-cell junctions and functions as a tumor suppressor in a wide range of carcinomas. MTSS1 binds phosphoinositide-rich membranes through its I-BAR domain and is capable of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service