Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA018826

Sigma-Aldrich

Anti-SLC43A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-L-type amino acid transporter 3, Anti-Large neutral amino acids transporter small subunit 3, Anti-Prostate cancer overexpressed gene 1 protein, Anti-Solute carrier family 43 member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NCTLNWPIEAFPAPEEVNYTKKIKLSGLALDHKVTGDLFYTHVTTMGQRLSQKAPSLEDGSDAFMSPQDVRGTSENLPERSVPLRKSLCS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC43A1(8501)

General description

The gene solute carrier family 43 member-1 (SLC43A1) is mapped to human chromosome 11p11.1-p11.2. It belongs to the SLC43 family. SLC43A1 transcripts are present in pancreas, liver, skeletal muscle, fetal liver, heart, placenta, lung, kidney, spleen, prostate, testis, ovary, small intestine, colon, lymph node and bone marrow. SLC43A1 is popularly called as LAT3 (L-type amino acid transporter 3).

Immunogen

Large neutral amino acids transporter small subunit 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 43 member-1 (SLC43A1) is a Na+-independent neutral amino acid transporter. In Xenopus oocytes, SLC43A1 transports neutral amino acids such as L-leucine, L-isoleucine, L-valine and L-phenylalanine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74762

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junryo Rii et al.
Cancer science, 112(9), 3871-3883 (2021-05-30)
L-type amino acid transporter 3 (LAT3, SLC43A1) is abundantly expressed in prostate cancer (PC) and is thought to play an essential role in PC progression through the cellular uptake of essential amino acids. Here, we analyzed the expression, function, and
Ellappan Babu et al.
The Journal of biological chemistry, 278(44), 43838-43845 (2003-08-22)
A cDNA that encodes a novel Na+-independent neutral amino acid transporter was isolated from FLC4 human hepatocarcinoma cells by expression cloning. When expressed in Xenopus oocytes, the encoded protein designated LAT3 (L-type amino acid transporter 3) transported neutral amino acids
Susanna Bodoy et al.
Molecular aspects of medicine, 34(2-3), 638-645 (2012-12-27)
The SLC43 family is composed of only three genes coding for the plasma membrane facilitator system l amino acid transporters LAT3 (SLC43A1; TC 2.A.1.44.1) and LAT4 (SLC43A2; TC 2.A.1.44.2), and the orphan protein EEG1 (SLC43A3; TC 2.A.1.44.3). Besides the known
K A Cole et al.
Genomics, 51(2), 282-287 (1998-09-02)
We recently identified a novel gene (PB39) (HGMW-approved symbol POV1) whose expression is up-regulated in human prostate cancer using tissue microdissection-based differential display analysis. In the present study we report the full-length sequencing of PB39 cDNA, genomic localization of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service