Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA017010

Sigma-Aldrich

Anti-PDLIM1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C- terminal LIM domain protein 1, Anti-Elfin, Anti-LIM domain protein CLP-36, Anti-PDZ and LIM domain protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

EKQELNEPPKQSTSFLVLQEILESEEKGDPNKPSGFRSVKAPVTKVAASIGNAQKLPMCDKCGTGIVGVFVKLRDRHRHPECYVCTDCGTNLKQKGHFFVEDQIYCEKHARERVTPPEGYEVVTV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM1(9124)

General description

PDLIM1 (PDZ and LIM domain 1) is a 36kDa carboxyl terminal LIM domain protein consisting of a PDZ domain at the amino terminal. It belongs to the enigma family of proteins. It is highly expressed in heart, skeletal muscles and in spleen, and at lower extent in small intestine, colon, placenta, liver, thymus, kidney, prostate, and pancreas. It is localized in the cytotrophoblast layer.

Immunogen

PDZ and LIM domain protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PDLIM1 (PDZ and LIM domain 1) acts as a cytoskeletal protein involved in the cytoskeleton reorganizations. It binds to α-actinin to control the cytoskeletal reorganization and signal transduction. It has been reported that PDLIM1 plays a key role in controlling the progression of breast cancer cells in terms of migration and metastasis control.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71113

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrich Miehe et al.
Histochemistry and cell biology, 126(4), 465-471 (2006-04-13)
Differentiation processes in the trophoblast comprise polarization, cell fusion and migration. All these processes involve dramatic reorganizations of cytoskeletal proteins such as intermediate filaments or actin. Due to very restricted knowledge on cytoskeletal changes in trophoblast, we analyzed the protein
M Kotaka et al.
Journal of cellular biochemistry, 72(2), 279-285 (1999-02-18)
We characterized a human cDNA clone encoding a 36-kDa carboxyl terminal LIM domain protein with a PDZ domain at the amino terminal. This full-length cDNA clone has a predicted open reading frame (ORF) of 329 amino-acid residues. The ORF of
Z Liu et al.
Oncogene, 34(10), 1300-1311 (2014-03-26)
Increased CLP36 expression has been found to be closely associated with breast cancer progression. However, whether and how it contributes to malignant behavior of breast cancer cells were not known. We show here that CLP36 is critical for promoting breast
Yixuan Lu et al.
Theranostics, 12(11), 5051-5068 (2022-07-16)
Background: p53 deficiency is a key causal factor for tumor development and progression. p53 acts in this process through, at least in part, cooperation with YAP1 but the underlying molecular mechanism is incompletely understood. In this paper, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service