Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA015955

Sigma-Aldrich

Anti-CTNND1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAS, Anti-Cadherin-associated Src substrate, Anti-Catenin delta-1, Anti-p120 catenin, Anti-p120(cas), Anti-p120(ctn

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SVKEQEAQFEKLTRALEEERRHVSAQLERVRVSPQDANPLMANGTLTRRHQNGRFVGDADLERQKFSDLKLNGPQDHSHLLYSTIPRMQEPGQIVETYTEEDPEGAMSVVSVETSDDGTTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNND1(1500)

Immunogen

Catenin delta-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Catenin δ-1 is a protein encoded by the CTNND1 gene in humans and belongs to p120 catenin family. It maintains cell-cell adhesion by binding to the juxtamembrane domain (JMD) of E-cadherin and may regulate Cdc42/Rac1 activity. This protein is involved in cancer progression thereby promoting proliferation and invasion of breast cancer cells, lung cancer and alters cell cycle progression. Overexpression of this protein is found in breast cancer and may act as a marker in oral squamous cell carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72653

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di Zhang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(2), 569-575 (2014-10-03)
δ-Catenin is a member of the p120 catenin family. Similar to p120ctn, δ-catenin contains nine central Armadillo repeats and binds to the juxtamembrane domain (JMD) of E-cadherin. We used immunohistochemistry to detect δ-catenin expression in breast carcinoma (128 cases), and
Lorenzo Lo Muzio et al.
Oral surgery, oral medicine, oral pathology and oral radiology, 115(6), 789-798 (2013-05-28)
p120ctn is a component of the catenin family. To date, there have only been two studies examining expression levels of p120ctn in oral squamous cell carcinoma (OSCC). Paraffined specimens of 113 OSCCs and 12 of normal mucosa were examined by
Zhangjun Cheng et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 32(5), 707-712 (2012-10-18)
This study examined the possible role of p120ctn in the pathogenesis and development of pancreatic cancer. PANC-1 cells, a kind of human pancreatic carcinoma cell line, were cultured in this study. p120ctn was immunocytochemically detected in PANC-1 cells. The recombinant
Mi Deng et al.
Journal of cellular physiology, 234(8), 13342-13352 (2019-01-17)
The activation and proliferation of human Tenon's fibroblasts (HTFs) play a vital role in the fibrosis in the pathology of the scar formation after the glaucoma filtration surgery. Transforming growth factor β1 (TGFβ1)/Smads signaling has been reported to promote fibrosis.
Wen-Ya Ma et al.
Acta pharmacologica Sinica, 42(6), 921-931 (2020-08-26)
The neonatal heart possesses the ability to proliferate and the capacity to regenerate after injury; however, the mechanisms underlying these processes are not fully understood. Melatonin has been shown to protect the heart against myocardial injury through mitigating oxidative stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service