Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

HPA014899

Sigma-Aldrich

Anti-ASGR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASGP-R 2, Anti-ASGPR 2, Anti-Asialoglycoprotein receptor 2, Anti-Hepatic lectin H2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:5000-1:10000

immunogen sequence

QSAQLQAELRSLKEAFSNFSSSTLTEVQAISTHGGSVGDKITSLGAKLEKQQQDLKADHDALLFHLKHFPVDLRFVACQMELLHSNGSQRTCC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASGR2(433)

General description

ASGR2 (asialoglycoprotein receptor 2) is one of the two subunits of the hetero-oligomeric ASGP-R, which is expressed specifically on hepatocytes. It is also called H2 and shares high homology with the other subunit called H1. ASGP-R is present on the plasma membrane of hepatocytes, towards the sinusoidal side, and undergoes endocytic recycling. This receptor is a type II membrane protein, and thus, ASGR2 has its N-terminal towards the cytoplasm, one transmembrane region, a stalk region and a Ca2+-dependent CRD (carbohydrate recognition domain). The molecular weight of this subunit is ~ 50kDa.

Immunogen

Asialoglycoprotein receptor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ASGR2 (asialoglycoprotein receptor 2) forms a subunit of ASGR, which internalizes desialylated glycoproteins, and transports them to lysosomes. It is also a receptor for hepatitis B virus (HBV), the chronic infection of which can affect the male reproductive system. As ASGR is also expressed in human testis, it might be responsible for HBV infection in the luminal compartment of testis. This receptor is also involved in the immune regulation and inflammatory processes in hepatocytes, and promotes liver cell toxicity. An alternatively spliced form of ASGR2 subunit, called sH2a is found in serum of healthy individuals. The serum level of this form is changed in liver fibrosis, and defines the status of hepatocyte function. Thus, it has potential as a non-invasive marker for liver function and fibrosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72295

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingnan Sun et al.
Cell and tissue research, 352(3), 761-768 (2013-04-23)
During acute or chronic hepatitis B virus (HBV) infection, the virus can invade the male reproductive system, pass through the blood-testis barrier and integrate into the germ line, resulting in abnormal spermatozoa. However, the pathway remains unclear. The asialoglycoprotein receptor
Elena Veselkin et al.
PloS one, 6(11), e27210-e27210 (2011-11-19)
The human asialoglycoprotein receptor is a membrane heterooligomer expressed exclusively in hepatocytes. A soluble secreted form, sH2a, arises, not by shedding at the cell surface, but by intracellular cleavage of its membrane-bound precursor, which is encoded by an alternatively spliced
Jasper H N Yik et al.
The Journal of biological chemistry, 277(25), 23076-23083 (2002-04-12)
The hepatic asialoglycoprotein receptor (ASGP-R) is an endocytic receptor that mediates the internalization of desialylated glycoproteins and their delivery to lysosomes. The human ASGP-R is a hetero-oligomeric complex composed of H1 and H2 subunits. There are three naturally occurring H2
Amit Saxena et al.
The Journal of biological chemistry, 277(38), 35297-35304 (2002-06-29)
The functional human hepatic asialoglycoprotein receptor (ASGP-R) is a hetero-oligomer composed of two subunits, designated H1 and H2, which are highly homologous. Despite their extensive homology, the major H1 subunit is stably expressed by itself, whereas in the absence of
Clifford S Guy et al.
Hepatology (Baltimore, Md.), 54(3), 1043-1050 (2011-06-10)
It has been recently identified that hepatocytes can act as cytotoxic effectors and can kill contacted cells by way of CD95 ligand-CD95 and perforin-dependent pathways. However, it remained unknown whether hepatocyte-mediated cell killing is indiscriminant or is directed toward targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service