Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA014343

Sigma-Aldrich

Anti-KLK5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KLK-L2, Anti-Kallikrein-5 precursor, Anti-Kallikrein-like protein 2, Anti-Stratum corneum tryptic enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

CDHPSNTVPSGSNQDLGAGAGEDARSDDSSSRIINGSDCDMHTQPWQAALLLRPNQLYCGAVLVHPQWLLTAAHCRKKVFRVRLGHYSLSPVYESGQQMFQGVKSIPHPGYSHPGHSNDLMLIKLNRRIRPTKDVRPINVSSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLK5(25818)

General description

The gene KLK5 (kallikrein related peptidase 5) encodes a serine protease that is a member of the kallikrein gene family. Its expression is abundant in skin, mammary gland and testis. The gene is localized to a gene cluster at human chromosome 19q13.2-q13.4. The encoded protein is synthesized as a preproenzyme containing an N-terminal signal peptide (prezymogen) followed by an activation peptide and the enzymatic domain. It is highly homologous to the enamel matrix serine proteinase 1 (EMSP1).

Immunogen

Kallikrein-5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-KLK5 antibody produced in rabbit has been used in immunohistochemistry (1.0 μg/mL).

Biochem/physiol Actions

The gene KLK5 (kallikrein related peptidase 5) encodes a serine protease that is also referred to as KLK-L2 or human stratum corneum tryptic enzyme that is involved in the degradation of corneodesmosome proteins at the epidermal surface. This is a necessary step in desquamation. The corneodesmosome proteins comprising of corneodesmosin (CDSN), desmoglein 1 (DSG1), and desmocollin 1 (DSC1) mediate corneocyte cohesion. KLK5 expression is linked to more aggressive forms of epithelial ovarian carcinoma and it may serve as an independent prognostic marker of poor prognosis in ovarian cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72196

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Meunier et al.
Journal of cosmetic dermatology, 18(4), 1140-1154 (2018-11-30)
Healthy skin is a delicate balance between skin renewal and microbiota homeostasis, and its imbalance promotes premature aging and dermatological disorders. Skin stem cells are key actors in this process but their sensitivity to aging and external stressors such as
H Kim et al.
British journal of cancer, 84(5), 643-650 (2001-03-10)
Kallikrein gene 5 (KLK5, also known as KLK-L2), located on chromosome 19q13.4, is one of the newly identified members of the kallikrein gene family, which is a subgroup of the serine protease enzyme family. In normal human tissues, KLK5 is
Cécile Caubet et al.
The Journal of investigative dermatology, 122(5), 1235-1244 (2004-05-14)
Corneodesmosin (CDSN), desmoglein 1 (DSG1), and desmocollin 1 (DSC1) are adhesive proteins of the extracellular part of the corneodesmosomes, the junctional structures that mediate corneocyte cohesion. The degradation of these proteins at the epidermis surface is necessary for desquamation. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service