Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

HPA014266

Sigma-Aldrich

Anti-GDAP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GDAP1, Anti-Ganglioside-induced differentiation-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PLSEHNEPWFMRLNSTGEVPVLIHGENIICEATQIIDYLEQTFLDERTPRLMPDKESMYY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GDAP1(54332)

General description

The gene GDAP1 (ganglioside induced differentiation associated protein 1) encodes a protein that is most abundantly expressed in neurons and to some extent in Schwann cells. It is found to be localized to mitochondria and contains two C-terminal transmembrane domains that are essential for correct localization in mitochondria. The encoded protein is 358 amino acid long having two characteristic GST (glutathione S-transferase) domains at the N-terminal region. Domain I serves as the glutathione binding site and domain II is related to sites linked to presumed cytotoxic and xenobiotic activities. The gene is mapped to human chromosome 8q21.11.

Immunogen

Ganglioside-induced differentiation-associated protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein ganglioside induced differentiation associated protein 1 has been associated with pathology of autosomal recessive (AR) demyelinating Charcot–Marie–Tooth disease (CMT) type 4A (CMT4A) as well as AR axonal CMT with vocal cord paralysis. It has also been implicated in autosomal dominant neuropathies. GDAP1 may participate in the maintenance of the mitochondrial network.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72835

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laia Pedrola et al.
Human molecular genetics, 14(8), 1087-1094 (2005-03-18)
Mutations in GDAP1, the ganglioside-induced differentiation-associated protein 1 gene, cause Charcot-Marie-Tooth (CMT) type 4A, a severe autosomal recessive form of neuropathy associated with either demyelinating or axonal phenotypes. Here, we demonstrate that GDAP1 has far greater expression in neurons than
E Nelis et al.
Neurology, 59(12), 1865-1872 (2002-12-25)
Mutations in the ganglioside-induced differentiation-associated protein 1 gene (GDAP1) were recently shown to be responsible for autosomal recessive (AR) demyelinating Charcot-Marie-Tooth disease (CMT) type 4A (CMT4A) as well as AR axonal CMT with vocal cord paralysis. The coding region of
Christina Wolf et al.
Communications biology, 5(1), 541-541 (2022-06-07)
Charcot-Marie-Tooth (CMT) disease 4A is an autosomal-recessive polyneuropathy caused by mutations of ganglioside-induced differentiation-associated protein 1 (GDAP1), a putative glutathione transferase, which affects mitochondrial shape and alters cellular Ca2+ homeostasis. Here, we identify the underlying mechanism. We found that patient-derived
Developmental Stage-Dependent Changes in Mitochondrial Function in the Brain of Offspring Following Prenatal Maternal Immune Activation.
Cie??lik, et al.
International Journal of Molecular Sciences, 24 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service