Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA013398

Sigma-Aldrich

Anti-ANXA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-35-alpha calcimedin, Anti-Annexin A3, Anti-Annexin III, Anti-Annexin-3, Anti-Inositol 1,2-cyclic phosphate 2-phosphohydrolase, Anti-Lipocortin III, Anti-PAP-III, Anti-Placental anticoagulant protein III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

ISQAYYTVYKKSLGDDISSETSGDFRKALLTLADGRRDESLKVDEHLAKQDAQILYKAGENRWGTDEDKFTEILCLRSFPQLKLTFDEYRNISQKDIVDSIKGELSGHFEDLLLAIVNCVRNTPAFLAERLHRALKGIGTD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA3(306)

Immunogen

Annexin A3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ANXA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ANXA3 (Annexin A3) gene encodes a calcium-dependent phospholipid-binding protein belonging to the annexin family. It inhibits phopholipase A2 and cleaves the cyclic bond of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. It functions as an anti-coagulant. It promotes migration and tube formation of human umbilical vein endothelial cells. It induces the production of vascular endothelial growth factor (VEGF) via the HIF-1 (hypoxia-inducible factor-1) pathway and functions as an angiogenic factor. Annexin A3 also inhibits adipocyte differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72660

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael L Merchant et al.
Journal of the American Society of Nephrology : JASN, 31(8), 1883-1904 (2020-06-21)
The mechanisms leading to extracellular matrix (ECM) replacement of areas of glomerular capillaries in histologic variants of FSGS are unknown. This study used proteomics to test the hypothesis that glomerular ECM composition in collapsing FSGS (cFSGS) differs from that of
Andrii Dinets et al.
PloS one, 10(5), e0126472-e0126472 (2015-05-16)
Cystic papillary thyroid carcinoma (cPTC) is a subgroup of PTC presenting a diagnostic challenge at fine needle aspiration biopsy (FNAB). To further investigate this entity we aimed to characterize protein profiles of cyst fluids from cPTC and benign thyroid cystic
Zengli Zhang et al.
Neural plasticity, 2021, 5575090-5575090 (2021-07-06)
The parenchymal microglia possess different morphological characteristics in cerebral physiological and pathological conditions; thus, visualizing these cells is useful as a means of further investigating parenchymal microglial function. Annexin A3 (ANXA3) is expressed in microglia, but it is unknown whether
Xiaofang Jia et al.
Proteome science, 10(1), 39-39 (2012-06-12)
Alcohol-induced injury has become one of the major causes for liver cirrhosis. However, the molecular mechanisms of ethanol-induced injury are not fully understood. To this end, we performed a dynamic plasma membrane proteomic research on rat model. A rat model
Takenori Watanabe et al.
Journal of biochemistry, 152(4), 355-363 (2012-08-14)
Annexin A3 is a protein belonging to the annexin family, and it is mainly present in cellular membranes as a phospholipid-binding protein that binds via the calcium ion. However, its physiological function remains to be clarified. We examined the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service