Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA012805

Sigma-Aldrich

Anti-PLXDC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Plexin domain-containing protein 1 precursor, Anti-Tumor endothelial marker 3, Anti-Tumor endothelial marker 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RSCDACMSSDLTFNCSWCHVLQRCSSGFDRYRQEWMDYGCAQEAEGRMCEDFQDEDHDSASPDTSFSPYDGDLTTTSSSLFIDSLTTEDDTKLNPYAGGDGLQNNLSPKTKGTPVHLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLXDC1(57125)

General description

Plexin domain-containing protein 1 (PLXDC1) is a transmembrane protein that is expressed in the tumor endothelium. It contains a signal peptide, a nidogen-like domain and a single hydrophobic domain.

Immunogen

Plexin domain-containing protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Plexin domain-containing protein 1 (PLXDC1) interacts with basement membrane protein, nidogen, on the cell surface. It is associated with metastasis of osteogenic sarcoma. PLXDC1 is up-regulated in the blood vessels of solid tumors of colon, lung and oesophageal cancers. It also has an important role in the proliferation of neovascular endothelial cells in the fibrovascular membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72123

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TEM7 (PLXDC1) in neovascular endothelial cells of fibrovascular membranes from patients with proliferative diabetic retinopathy.
Yamaji Y, et al.
Investigative Ophthalmology & Visual Science, 49(7), 3151-3157 (2008)
Identification of the basement membrane protein nidogen as a candidate ligand for tumor endothelial marker 7 in vitro and in vivo.
Lee HK, et al.
Febs Letters, 580(9), 2253-2257 (2006)
Bruno Fuchs et al.
Gene, 399(2), 137-143 (2007-06-15)
Our objective is to identify genes regulating metastasis of osteogenic sarcoma (OGS) since metastasis is the primary cause of mortality among patients with OGS. To identify such genes, we first created a database of differentially expressed genes between six low-grade
Akash Nanda et al.
Cancer research, 64(23), 8507-8511 (2004-12-03)
Tumor endothelial marker 7 (TEM7) was recently identified as an mRNA transcript overexpressed in the blood vessels of human solid tumors. Here, we identify several new variants of TEM7, derived by alternative splicing, that are predicted to be intracellular (TEM7-I)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service