Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

HPA012783

Sigma-Aldrich

Anti-DPEP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dehydropeptidase-I, Anti-Dipeptidase 1 precursor, Anti-Microsomal dipeptidase, Anti-Renal dipeptidase, Anti-hRDP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500
western blot: 0.04-0.4 μg/mL

immunogen sequence

SLIGVEGGHSIDSSLGVLRALYQLGMRYLTLTHSCNTPWADNWLVDTGDSEPQSQGLSPFGQRVVKELNRLGVLIDLAHVSVATMKATLQLSRAPVIFSHSSAYSVCASRRN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPEP1(1800)

General description

Dipeptidase 1 (DPEP1) is present in the kidney membrane. It contains 411 residues and a signal peptide of 16 residues is located at the carboxyl terminus. The gene encoding this protein is located on chromosome 16q24.3.

Immunogen

Dipeptidase 1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-DPEP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Dipeptidase 1 (DPEP1) hydrolyzes a variety of dipeptides and is highly expressed in colon tumors. It functions in the renal metabolism of glutathione and its conjugates, and also hydrolyzes β-lactam antibiotics. DPEP1 expression in cells and tissues inhibits tumour cell invasiveness in pancreatic ductal adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71250

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazunoshin Tachibana et al.
Biomedical reports, 6(4), 423-428 (2017-04-18)
Dipeptidase 1 (DPEP1) is a zinc-dependent metalloproteinase that is fundamental in glutathione and leukotriene metabolism.
P A Eisenach et al.
British journal of cancer, 109(3), 694-703 (2013-07-11)
Colorectal cancer (CRC) is the second leading cause of cancer-related deaths worldwide. Improvements in the understanding of its molecular mechanism and the characterisation of CRC-specific biomarkers facilitating early detection are considered to increase overall survival. A meta-analysis of microarray and
E Austruy et al.
Genomics, 15(3), 684-687 (1993-03-01)
We report the subregional physical and genetic mapping on chromosome 16q of a cDNA clone selected as a potential tumor/growth suppressor sequence. By DNA sequencing and RNA expression pattern, this clone was identified as part of the renal dipeptidase gene
Takako Okamoto et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(2), 267-276 (2010-11-16)
Distinguishing primary mucinous ovarian cancers from ovarian metastases of digestive organ cancers is often challenging. Dipeptidase 1 was selected as the candidate novel marker of colorectal cancer based on an analysis of a gene expression microarray. Immunohistochemical analysis indicated that
H Adachi et al.
The Journal of biological chemistry, 265(7), 3992-3995 (1990-03-05)
Two cDNA clones corresponding to human microsomal dipeptidase (MDP, formerly referred to as dehydropeptidase-I or renal dipeptidase (EC 3.4.13.11] were isolated from human placental and renal cDNA libraries employing rapid amplification of cDNA ends strategy. The complete amino acid sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service