Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA011422

Sigma-Aldrich

Anti-SGCB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-43 kDa dystrophin-associated glycoprotein, Anti-43DAG, Anti-A3b, Anti-Beta-SG, Anti-Beta-sarcoglycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

FHMGGNMELKAENSIILNGSVMVSTTRLPSSSSGDQLGSGDWVRYKLCMCADGTLFKVQVTSQNMGCQISDNPCGNTH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGCB(6443)

Immunogen

Beta-sarcoglycan recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SGCB (Sarcoglycan, β) is a 43kD glycosylated transmembrane, dystrophin-associated glycoprotein. It is ubiquitously expressed, but mainly in skeletal and cardiac muscle. It forms and maintains the assembly and functional stability of the sarcoglycan complex by forming disulfide-bond. The complex includes α, β, γ, and δ, which are tightly bound to each other. The mutation in the sarcoglycan complex is associated with four types of autosomal recessive limb-girdle muscular dystrophies (LGMD2C-F).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72131

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara F Henriques et al.
PloS one, 13(1), e0191274-e0191274 (2018-01-24)
Sarcoglycanopathies are rare autosomic limb girdle muscular dystrophies caused by mutations in one of the genes coding for sarcoglycan (α, β, δ, and γ-sarcoglycans). Sarcoglycans form a complex, which is an important part of the dystrophin-associated glycoprotein complex that protects
R Barresi et al.
Journal of medical genetics, 37(2), 102-107 (2000-02-09)
Two young males with limb-girdle muscular dystrophy (LGMD) resulting from sarcoglycan deficiency died at 27 (patient 1) and 18 years (patient 2) of severe cardiomyopathy. Genetic analysis showed that they were compound heterozygotes for mutations in the beta sarcoglycan gene.
C G Bönnemann et al.
Neuromuscular disorders : NMD, 8(3-4), 193-197 (1998-06-19)
Four of the currently recognized autosomal recessive limb-girdle muscular dystrophies (LGMD type 2C-F) are caused by mutations in the genes encoding components of the sarcoglycan complex. LGMD 2C, caused by mutations in gamma-sarcoglycan, is prevalent in northern Africa, especially in
F Duclos et al.
Neuromuscular disorders : NMD, 8(1), 30-38 (1998-05-05)
The sarcoglycan complex is involved in the etiology of four autosomal recessive limb-girdle muscular dystrophies (LGMD2C-F). A missense mutation (T151R) in the beta-sarcoglycan gene on chromosome 4q12 has been shown to cause a mild form of LGMD2E in 11 families

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service