Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

HPA011316

Sigma-Aldrich

Anti-ACSL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ACS1, Anti-Acyl- CoA synthetase 1, Anti-LACS 1, Anti-LACS 2, Anti-Long-chain acyl-CoA synthetase 1, Anti-Long-chain acyl-CoA synthetase 2, Anti-Long-chain fatty acid-CoA ligase 2, Anti-Long-chain-fatty-acid--CoA ligase 1, Anti-Palmitoyl-CoA ligase 1, Anti-Palmitoyl-CoA ligase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

YATRPKPLKPPCDLSMQSVEVAGSGGARRSALLDSDEPLVYFYDDVTTLYEGFQRGIQVSNNGPCLGSRKPDQPYEWLSYKQVAELSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACSL1(2180)

General description

ACSL1 (acyl-CoA synthetase long-chain family member 1) is the main isoform of the long-chain acyl coenzyme A (acyl-CoA) synthetase (ACSL) isoenzymes. It is predominantly expressed in tissues with high energy metabolism such as, fat, skeletal muscles and liver. This gene maps to human chromosome 4q34-q35.

Immunogen

Long-chain-fatty-acid--CoA ligase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ACSL1 (acyl-CoA synthetase long-chain family member 1) is involved in the synthesis of triacylglycerols (TAG) and fatty acid metabolism. It activates and traps free fatty acids (FFAs) within cells. It catalyzes the conversion of FFAs to acyl coenzyme A (acyl-CoA) in the presence of ATP. It also has a role in heart metabolism. This gene has a putative role in determining endurance performance, and SNP rs6552828 is marginally linked with elite endurance performance in Chinese (Han) male population. However, no such association is found in Chinese female or Caucasian population. Polymorphism rs9997745 in ACSL1 is linked with the risk of metabolic syndrome. Eicosapentaenoic acid down-regulates the expression of this gene, which in turn leads to the down-regulation of palmitate-induced cytokine production. This results in the decrease of chronic inflammation in atherosclerotic plaques.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71196

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Yvert et al.
PloS one, 7(7), e41268-e41268 (2012-07-26)
The aim of this study was to determine the association between the rs6552828 polymorphism in acyl coenzyme A synthetase (ACSL1) and elite endurance athletic status. We studied 82 Caucasian (Spanish) World/Olympic-class endurance male athletes, and a group of sex and
The human palmitoyl-CoA ligase (FACL2) gene maps to the chromosome 4q34-q35 region by fluorescence in situ hybridization (FISH) and somatic cell hybrid panels.
E S Cantú et al.
Genomics, 28(3), 600-602 (1995-08-10)
Catherine M Phillips et al.
Journal of lipid research, 51(7), 1793-1800 (2010-02-24)
Long-chain acyl CoA synthetase 1 (ACSL1) plays an important role in fatty acid metabolism and triacylglycerol (TAG) synthesis. Disturbance of these pathways may result in dyslipidemia and insulin resistance, hallmarks of the metabolic syndrome (MetS). Dietary fat is a key
Masanori Nakakuki et al.
Atherosclerosis, 227(2), 289-296 (2013-02-26)
Chronic inflammation caused by macrophages may be associated with progression of arteriosclerosis or obesity, both risk factors for cardiovascular events. In the Japan EPA Lipid Intervention Study (JELIS), eicosapentaenoic acid (EPA), an n-3 polyunsaturated fatty acid, was found to reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service