Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA010919

Sigma-Aldrich

Anti-MGAT5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GNT-V, Anti-KIAA0773, Anti-mannosyl (α-1,6-)-glycoprotein β-1,6-N-acetyl-glucosaminyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ALQVKLAEPGQSCKQVCQESQLICEPSFFQHLNKDKDMLKYKVTCQSSELAKDILVPSFDPKNKHCVFQGDLLLFSCAGAHPRHQRVCPCRDFIKGQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MGAT5(4249)

General description

MGAT5 (mannosyl (α-1,6-)-glycoprotein β-1,6-N-acetyl-glucosaminyltransferase) is a glycosyltransferase, which resides in the golgi apparatus. This gene maps to human chromosome 2q21 and has an open reading frame (ORF) of 2349bp. It is expressed ubiquitously, with the exception of skeletal muscle. Also its expression levels in brain are low.

Immunogen

mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MGAT5 (mannosyl (α-1,6-)-glycoprotein β-1,6-N-acetyl-glucosaminyltransferase) catalyzes the addition of β1-6 N-acetylglucosamine branches on asparagine (N)-linked oligosaccharides (N-glycan), present on cell proteins. It catalyzes the transfer of N-acetylglucosamine (GlcNAc) to β1,6-linked mannosyl residue, at their C-6 position, within the trimannosyl core structure of N-glycans, which are complex in nature. This produces GlcNAc(β1,6)[GlcNAcβ1,2] mannose (Man) (α1,6). Dysregulation of this enzyme is associated with many cancers. The expression of MGAT5 in gastric cancer is related to metastasis, tumor grade and stage and prognosis. It is also down-regulated in ulcerative colitis (UC), which leads to disruption of N-glycosylation of T-cell receptor. Hence, MGAT5 plays a role in the pathogenesis of UC. The expression of MGAT5 is positively correlated with proliferation and metastasis of human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72105

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana M Dias et al.
Human molecular genetics, 23(9), 2416-2427 (2013-12-18)
The incidence of inflammatory bowel disease is increasing worldwide and the underlying molecular mechanisms are far from being fully elucidated. Herein, we evaluated the role of N-glycosylation dysregulation in T cells as a key mechanism in the ulcerative colitis (UC)
Mika Kaneko et al.
FEBS letters, 554(3), 515-519 (2003-11-19)
UDP-N-acetylglucosamine:alpha(1,6)-D-mannoside beta(1,6)-N-acetylglucosaminyltransferase (GnT-V, Mgat5) functions in the biosynthesis of N-linked glycans and is transcriptionally upregulated by oncogene signaling. We report here the cloning and characterization of a human cDNA encoding a distinct enzyme with related substrate specificity, termed GnT-VB, which
Ting Wei et al.
Experimental and molecular pathology, 93(1), 8-17 (2012-04-28)
To investigate the role of N-acetylglucosaminyltransferases V (GnT-V) in the malignancy of human hepatocellular carcinoma (HCC), the GnT-V stably suppressed cell line HepG2 GnT-V/1564 was constructed from HepG2. The proliferation, migration, invasion, metastasis of HepG2 GnT-V/1564 was investigated both in
Binbin Huang et al.
International journal of oncology, 44(3), 849-857 (2014-01-09)
N-acetylglucosaminyltransferase V (GnT-V) is an enzyme that catalyzes β1-6 branching of N-acetylglucosamine on asparagine (N)-linked oligosaccharides (N-glycan) of cell proteins and the dysfunction of which is a common feature of various carcinomas. Nevertheless, the role of GnT-V remains controversial. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service