Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA009413

Sigma-Aldrich

Anti-JPH1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-JP-1 antibody produced in rabbit, Anti-Junctophilin type 1 antibody produced in rabbit, Anti-Junctophilin-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PQSKYSGRHHIPNPSNGELHSQYHGYYVKLNAPQHPPVDVEDGDGSSQSSSALVHKPSANKWSPSKSVTKPVAKESKAEPKAKKSELAIPKNPASNDSCPAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JPH1(56704)

Looking for similar products? Visit Product Comparison Guide

General description

JPH1 (junctophilin 1) is a subtype of the junctional membrane complex protein called JP, which contains three subtypes. This subtype is predominantly expressed in skeletal muscles. It is expressed in the junctional membrane complexes of excitable cells, and exists as a transmembrane protein. This gene is localized to human chromosome 8q21.1.

Immunogen

Junctophilin-1 recombinant protein epitope signature tag (PrEST)

Application

Anti-JPH1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Junctophilins span the membrane of intracellular Ca2+ storage vesicles and aids in their interaction with cell-surface membrane, thus, facilitating the formation of junctional membrane complexes. Studies in mice show that this protein plays a crucial role in the formation of triads during skeletal muscle development. JPH1 and GADP-1 are parts of a common pathway, and hence, interdependent. Thus, mutations in GDAP-1 (ganglioside-induced differentiation-associated protein 1) might phenotypically be manifested by JPH1. Therefore, JPH1 is implicated in Charcot-Marie-Tooth (CMT) disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71724

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nishi et al.
Biochemical and biophysical research communications, 273(3), 920-927 (2000-07-13)
Junctophilin (JP) subtypes, namely JP-1, 2, and 3, have been currently identified in excitable cells and constitute a novel family of junctional membrane complex proteins. Our studies have suggested that JPs take part in the formation of junctional membrane complexes
Shinji Komazaki et al.
FEBS letters, 524(1-3), 225-229 (2002-07-24)
Junctophilins (JP-1, JP-2, and JP-3) are transmembrane proteins expressed in the junctional membrane complexes in excitable cells. Both JP-1 and JP-2 are co-expressed in the triads of skeletal muscle, but only JP-2 is expressed in cardiac muscle. We analyzed the
Simona Boncompagni et al.
PloS one, 7(7), e39962-e39962 (2012-07-07)
Triadin (Tdn) and Junctin (Jct) are structurally related transmembrane proteins thought to be key mediators of structural and functional interactions between calsequestrin (CASQ) and ryanodine receptor (RyRs) at the junctional sarcoplasmic reticulum (jSR). However, the specific contribution of each protein
David Pla-Martín et al.
Human molecular genetics, 24(1), 213-229 (2014-08-30)
Mutations in the GDAP1 gene cause different forms of Charcot-Marie-Tooth (CMT) disease, and the primary clinical expression of this disease is markedly variable in the dominant inheritance form (CMT type 2K; CMT2K), in which carriers of the GDAP1 p.R120W mutation
Cecilia Lindskog et al.
BMC genomics, 16, 475-475 (2015-06-26)
To understand cardiac and skeletal muscle function, it is important to define and explore their molecular constituents and also to identify similarities and differences in the gene expression in these two different striated muscle tissues. Here, we have investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service