Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA008482

Sigma-Aldrich

Anti-CA14 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CA-XIV antibody produced in rabbit, Anti-Carbonate dehydratase XIV antibody produced in rabbit, Anti-Carbonic anhydrase 14 precursor antibody produced in rabbit, Anti-Carbonic anhydrase XIV antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GQHWTYEGPHGQDHWPASYPECGNNAQSPIDIQTDSVTFDPDLPALQPHGYDQPGTEPLDLHNNGHTVQLSLPSTLYLGGLPRKYVAAQLHLHWGQKGSPGGSEHQINSEATFAELHIVHYDSDSYDSLSEAAERPQGLAVLGIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CA14(23632)

General description

Carbonic anhydrase 14 (CA14) is a metalloenzyme, which is localized in the brain. It belongs to the α-CA gene family and the gene encoding it is present on the chromosome 1q21. It has a hydrophobic N-terminal signal sequence and a transmembrane domain at the C-terminal.

Immunogen

Carbonic anhydrase 14 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Carbonic anhydrase 14 (CA14) catalyzes the reversible hydration of carbon dioxide. It is also involved in modulating the response to excitatory stimuli at synaptic termini. CA14 regulates intracellular pH in hippocampal neurons by regulating AE3-mediated Cl-–HCO3- exchange.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71841

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Purkerson et al.
Kidney international, 71(2), 103-115 (2006-12-14)
Carbonic anhydrase (CA) catalyzes the reversible hydration of CO(2). CA is expressed in most segments of the kidney. CAII and CAIV predominate in human and rabbit kidneys; in rodent kidneys, CAXII, and CAXIV are also present. CAIX is expressed by
S Parkkila et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(4), 1918-1923 (2001-02-15)
Although long suspected from histochemical evidence for carbonic anhydrase (CA) activity on neurons and observations that CA inhibitors enhance the extracellular alkaline shifts associated with synaptic transmission, an extracellular CA in brain had not been identified. A candidate for this
Nataliya Svichar et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(10), 3252-3258 (2009-03-13)
Carbonic anhydrase (CA) activity in the brain extracellular space is attributable mainly to isoforms CA4 and CA14. In brain, these enzymes have been studied mostly in the context of buffering activity-dependent extracellular pH transients. Yet evidence from others has suggested
Claudia Temperini et al.
Bioorganic & medicinal chemistry letters, 17(3), 628-635 (2006-11-28)
The activation of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1) with L-adrenaline and histamine has been investigated by kinetic and X-ray crystallographic studies. L-Adrenaline behaves as a potent activator of isozyme CA I (activation constant of 90 nM), being a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service