Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA007613

Sigma-Aldrich

Anti-GALNT3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GalNAc-T3 antibody produced in rabbit, Anti-Polypeptide GalNAc transferase 3 antibody produced in rabbit, Anti-Polypeptide N-acetylgalactosaminyltransferase 3 antibody produced in rabbit, Anti-Protein-UDP acetylgalactosaminyltransferase 3 antibody produced in rabbit, Anti-UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3 antibody produced in rabbit, Anti-pp-GaNTase 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EESRMERNMKNKNKMLDLMLEAVNNIKDAMPKMQIGAPVRQNIDAGERPCLQGYYTAAELKPVLDRPPQDSNAPGASGKAFKTTNLSVEEQKEKERGEAKHC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALNT3(2591)

General description

GALNT3 (polypeptide N-acetylgalactosaminyltransferase 3) is a cell adhesion molecule belonging to the polypeptide N-acetylgalactosaminyltransferases (GalNAc-Ts) family. It is majorly involved in the regulation of initial steps of mucin-type O-glycosylation.

Immunogen

polypeptide N-acetylgalactosaminyltransferase 3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GALNT3 (polypeptide N-acetylgalactosaminyltransferase 3) gene encoded proteins catalyze the transfer of GalNAc from UDP-GalNAc to either serine or threonine residues of polypeptide acceptors. GALNT3 catalyzes the initiation of O-linked oligosaccharide biosynthesis. Protein glycosylation of target proteins is important in stabilizing the half-life, biological activity and receptor recognition of these proteins. Defects in this gene have been associated with rare autosomal recessive metabolic disorder familial tumoral calcinosis characterized by hyperphosphatemia and massive ectopic calcifications.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71818

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kitada et al.
British journal of cancer, 109(2), 472-481 (2013-06-27)
The polypeptide N-acetylgalactosaminyltransferases (GalNAc-Ts) family of enzymes regulates the initial steps of mucin-type O-glycosylation. N-acetylgalactosaminyltransferases might show novel patterns of GalNAc-T glycosylation on tumour-derived proteins, which could influence cancer biology, but its mechanisms are unclear. We investigated the association of
Shengjun Wang et al.
Nature communications, 15(1), 4162-4162 (2024-05-17)
The multibasic furin cleavage site at the S1/S2 boundary of the spike protein is a hallmark of SARS-CoV-2 and plays a crucial role in viral infection. However, the mechanism underlying furin activation and its regulation remain poorly understood. Here, we
E P Bennett et al.
The Journal of biological chemistry, 271(29), 17006-17012 (1996-07-19)
The glycosylation of serine and threonine residues during mucin-type O-linked protein glycosylation is carried out by a family of UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferases (GalNAc-transferase). Previously two members, GalNAc-T1 and -T2, have been isolated and the genes cloned and characterized. Here we report
Kentaro Kato et al.
The Journal of biological chemistry, 281(27), 18370-18377 (2006-04-28)
Mutations in the gene encoding the glycosyltransferase polypeptide GalNAc-T3, which is involved in initiation of O-glycosylation, were recently identified as a cause of the rare autosomal recessive metabolic disorder familial tumoral calcinosis (OMIM 211900). Familial tumoral calcinosis is associated with
Virginia Lorenz et al.
Biological chemistry, 398(11), 1237-1246 (2017-07-05)
O-GalNAc glycans are important structures in cellular homeostasis. Their biosynthesis is initiated by members of the polypeptide GalNAc-transferase (ppGalNAc-T) enzyme family. Mutations in ppGalNAc-T3 isoform cause diseases (congenital disorders of glycosylation) in humans. The K626 residue located in the C-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service