Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA006997

Sigma-Aldrich

Anti-S100A7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein S100-A7 antibody produced in rabbit, Anti-Psoriasin antibody produced in rabbit, Anti-S100 calcium-binding protein A7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

EKPSLLTMMKENFPNFLSACDKKGTNYLADVFEKKDKNEDKKIDFSEFLSLLGDIATDYHKQSHGAAPCS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A7(6278)

General description

S100A7 (S100 calcium binding protein A7) is Ca2+-binding protein associated with the various cellular and extracellular activities. It is expressed in the ear, skin, tongue and stratified squamous epithelial tissues.

Immunogen

Protein S100-A7 recombinant protein epitope signature tag (PrEST)

Application

Anti-S100A7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

S100A7 (S100 calcium binding protein A7) controls the epidermal growth factor (EGF)-induced cell migration and the actin remodeling. It has been suggested that S100A7 may stimulate metastasis of ERα(-) breast cancer cells. Overexpression of S100A7 gene causes inflammatory stimuli.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70732

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amita Sneh et al.
Breast cancer research and treatment, 138(3), 727-739 (2013-03-29)
Psoriasin (S100A7) is a calcium-binding protein that has shown to be highly expressed in high-grade ductal carcinoma in situ (DCIS) and a subset of invasive breast cancers. However, its role in invasion and metastasis is not very well known. In
D M Bürgisser et al.
Biochemical and biophysical research communications, 217(1), 257-263 (1995-12-05)
The Ca(2+)-binding proteins regulate a number of cellular and extracellular activities and deregulations of S100 gene expression are associated with several human diseases. For example, S100A7 is upregulated in psoriatic skin, implicating a link with psoriasis, a chronic inflammatory dermatosis.
Claire Jardet et al.
Experimental dermatology, 29(10), 993-1003 (2020-08-02)
Skin models mimicking features of psoriasis-related inflammation are needed to support the development of new drugs in dermatology. Reconstructed skin models lack tissue complexity, including a fully competent skin barrier, and presence and/or diversity of immune cells. Here, we describe
P Madsen et al.
The Journal of investigative dermatology, 97(4), 701-712 (1991-10-01)
Analysis of the protein patterns of normal and psoriatic noncultured unfractionated keratinocytes has revealed several low-molecular-weight proteins that are highly up-regulated in psoriatic epidermis. Here, we have cloned and sequenced the cDNA (clone 1085) for one of these proteins that
Joanna Y Lee et al.
Nature communications, 10(1), 1848-1848 (2019-04-25)
Increased tissue stiffness is a driver of breast cancer progression. The transcriptional regulator YAP is considered a universal mechanotransducer, based largely on 2D culture studies. However, the role of YAP during in vivo breast cancer remains unclear. Here, we find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service