Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA005673

Sigma-Aldrich

Anti-TRIM21 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RNF81, Anti-RO52, Anti-Ro/SSA, Anti-SSA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LEKDEREQLRILGEKEAKLAQQSQALQELISELDRRCHSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM21(6737)

General description

Tripartite motif containing 21 (TRIM21) belongs to the TRIM family of proteins which function as anti-retroviral restriction factors and are E3 ubiquitin-protein ligases.

Immunogen

52 kDa Ro protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tripartite motif containing 21 (TRIM21) is involved in regulation of IgG functions and has a role in innate immunity. It binds specifically to the Fc region of human IgG isotypes 1, 2 and 4 with a conformation dependent interaction. The minimal binding epitope is the C-terminal B30.2 domain. TRIM21 forms a trimer that competes with protein A for binding to IgG Fc and binds to the consensus CH2/CH3 domain interface in the Fc region, overlapping the binding site of several other proteins, like protein A and protein G. It has been shown that it interacts with interferon regulatory factor 3 (IRF3) upon RNA virus infection. TRIM21 interferes with the interaction between Pin1 (peptidyl-prolyl cis/trans isomerase, NIMA-interacting 1) and IRF3, thus preventing IRF3 ubiquitination and degradation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70028

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Yang et al.
Journal of immunology (Baltimore, Md. : 1950), 182(6), 3782-3792 (2009-03-07)
Virus infection induces host antiviral responses including induction of type I IFNs. Transcription factor IFN regulatory factor 3 (IRF3) plays an essential role and is tightly regulated in this process. Herein we report that TRIM21 (tripartite motif-containing 21) is significantly
David A Rhodes et al.
Molecular immunology, 44(9), 2406-2414 (2006-11-23)
TRIMs comprise a large protein family that include anti-retroviral restriction factors such as TRIM5alpha. Auto-antibodies to TRIM21 (Ro52) are a common serological feature of patients with Sjogren's syndrome and systemic lupus erythematosus (SLE). We show that, in addition to this
Keiji Wada et al.
Biochemical and biophysical research communications, 339(1), 415-421 (2005-11-22)
Anti-Ro/SSA antibodies are classic autoantibodies commonly found in patients with Sjögren's syndrome, a chronic autoimmune disease characterized by dryness of the eyes and mouth. The autoantibodies recognize a RING-finger protein, Ro52, whose function is still unknown. Since many RING-finger proteins
Cuiwei Yang et al.
Autophagy, 19(7), 1934-1951 (2023-01-25)
Eukaryotic stress granules (SGs) are highly dynamic assemblies of untranslated mRNAs and proteins that form through liquid-liquid phase separation (LLPS) under cellular stress. SG formation and elimination process is a conserved cellular strategy to promote cell survival, although the precise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service