Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA005449

Sigma-Aldrich

Anti-SLC2A5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fructose transporter antibody produced in rabbit, Anti-Glucose transporter type 5, small intestine antibody produced in rabbit, Anti-Solute carrier family 2, facilitated glucose transporter member 5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KALQTLRGWDSVDREVAEIRQEDEAEKAAGFISVLKLFRMRSLRWQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A5(6518)

General description

Glucose transporters (GLUTs) are a family of transporters that have 12 isoforms. These isoforms differ in function, tissue distribution and developmental regulation. Solute carrier family 2, member 5 (SLC2A5), also called GLUT5 is one such isoform, which is expressed in kidney, sperm cells and small intestine. It is the only transporter in the GLUT family, which is specific for fructose and is incapable of transporting glucose or galactose. The human SLC2A5 gene encodes for a 501 amino acid protein and has 12 exons. In humans, this gene is localized to the short arm of chromosome 1.

Immunogen

Solute carrier family 2, facilitated glucose transporter member 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 2, member 5 (SLC2A5) is responsible for the transport of fructose in the lumen of small intestine, kidney, brain and other insulin responsive tissues such as adipose tissue and muscle. It might also be responsible for recapturing the fructose lost in glomerular filtration. The fructose uptake rate and the expression levels of SLC2A5 depend upon various conditions such as obesity, diabetes, hypertension and inflammation (metabolic syndrome). Carcinogenesis has been shown to induce SLC2A5 expression, especially in mammary glands.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86879

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Wasserman et al.
The Journal of clinical investigation, 98(10), 2398-2402 (1996-11-15)
Fructose, a naturally occurring monosaccharide, is increasingly used as an added sweetener in processed foods in the form of high fructose corn syrup. Increased fructose intake combined with the identification of children with clinical evidence of isolated fructose malabsorption (IFM)
V Medina Villaamil et al.
Oncology reports, 25(2), 315-323 (2010-12-18)
Renal cell carcinomas (RCC) can be subclassified for general purposes into clear cell, papillary cell, chromophobe cell carcinomas and oncocytomas. Other tumours such as collecting duct, medullary, mucinous tubular and spindle cell and associated with Xp 11.2 translocations/TFE 3 gene
L Mahraoui et al.
The Biochemical journal, 301 ( Pt 1), 169-175 (1994-07-01)
The effect of cyclic AMP on the expression of the fructose transporter, GLUT5, was studied in Caco-2 cells, a human colon cancer cell line that differentiates spontaneously in culture into cells with the properties of small intestine enterocytes. Treatment of
T Kayano et al.
The Journal of biological chemistry, 265(22), 13276-13282 (1990-08-05)
Two novel facilitative glucose transporter-like cDNAs have been isolated from human small intestine and fetal skeletal muscle cDNA libraries by low stringency cross-hybridization with a fragment of the human erythrocyte/GLUT1 facilitative glucose transporter cDNA. One encodes a 501-amino acid facilitative
Shotaro Hayashida et al.
Brain pathology (Zurich, Switzerland), 30(6), 1144-1157 (2020-09-10)
TMEM119 and purinergic receptor P2Y12 (P2RY12), which are not expressed by recruited peripheral blood macrophages, are proposed to discriminate microglia from macrophages. Therefore, we investigated the distribution patterns of microglia and macrophages in 10 concentric lesions from four autopsied Baló's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service