Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA004098

Sigma-Aldrich

Anti-ZIC1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein ZIC 1, Anti-Zinc finger protein of the cerebellum 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LGINPFADGMGAFKLNPSSHELASAGQTAFTSQAPGYAAAAALGHHHHPGHVGSYSSAAFNSTRDFLFRNRGFGDAAAAASAQHSLFAASAGGFGGPHGHTDAAGHLLFPGLHEQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZIC1(7545)

Immunogen

Zinc finger protein ZIC 1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

ZIC1 is a member of the Zinc finger of the cerebellum (ZIC) protein family and is referred as ZIC and ZNF201. The gene encodes zinc finger containing proteins that can bind proteins and DNA. It is a vital transcription factor with zinc finger domains implicated in the process of neural development. It is a novel tumor suppressor gene that plays a crucial role in vertebrate development. Methylation of this gene may be a promising biomarker for the diagnosis and prognosis of HCC and ovarian cancer (OC). It serves as a potential therapeutic target for gastric cancer. It is also involved in progression of human cancers, including medulloblastomas, endometrial cancers and mesenchymal neoplasms. It acts as a novel candidate tumor suppressor gene down-regulated through promoter hypermethylation in gastric cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86720

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui-Lan Huang et al.
Epigenetics, 8(6), 624-634 (2013-06-19)
Women with advanced stage ovarian cancer (OC) have a five-year survival rate of less than 25%. OC progression is associated with accumulation of epigenetic alterations and aberrant DNA methylation in gene promoters acts as an inactivating "hit" during OC initiation
Yan-Ying Wang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(8), 7429-7433 (2014-05-02)
Hepatocellular carcinoma (HCC) is one of the most aggressive malignancies in humans, and its prognosis is generally poor even after surgery. The zinc finger of the cerebellum (ZIC1) gene is a novel tumor suppressor gene that plays a crucial role
Jing Zhong et al.
BMC cancer, 12, 290-290 (2012-07-18)
ZIC1, a vital transcription factor with zinc finger domains, has been implicated in the process of neural development. We previously showed that ZIC1 may function as a tumour suppressor in gastrointestinal cancers. However, the molecular mechanism underlying ZIC1 participation in
Dwayne B Holmes et al.
Biology open, 6(3), 402-406 (2017-02-09)
Here, we report a 3D cerebellar differentiation protocol with quick startup method, defined medium and no special materials or handling requirements. Three fibroblast growth factors (FGF2, 4 and 8) were used for cerebellar patterning and smoothened agonist (SAG) for granule
Radiya G Ali et al.
The international journal of biochemistry & cell biology, 44(11), 2065-2068 (2012-09-12)
The Zic genes encode zinc finger containing proteins that can bind proteins and DNA. The understanding of Zic molecular networks has been hampered by functional redundancy amongst family members, and because their loss-of-function phenotypes are indicative of a role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service