Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA003425

Sigma-Aldrich

Anti-TFF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Breast cancer estrogen-inducible protein, Anti-HP1A, Anti-PNR-2, Anti-Trefoil factor 1 precursor, Anti-pS2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

QTETCTVAPRERQNCGFPGVTPSQCANKGCCFDDTVRGVPWCFYPNTIDVPPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFF1(7031)

Immunogen

Trefoil factor 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Trefoil factor 1 is a protein encoded by the TFF1 gene in humans. The proteins are small secretory proteins characterized by three intra-chain disulfide bonds forming the trefoil motif or P-domain. It is abundantly expressed on mucosal surfaces and especially within the gastrointestinal tract. The protein belongs to highly conserved proteins that play an essential role in epithelial regeneration within the gastrointestinal tract. Its expression is strongly induced after mucosal injury and may function as a gastric tumor suppressor gene. It is expressed in various types of carcinomas including breast cancer. The gene contains an estrogen response element and its expression can be regulated by estrogen. Increased expression of this gene is found to be associated with tumor progression in some tumors and missense mutations in the gene have been detected in gastric cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86373

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arka Saha et al.
Cancers, 14(18) (2022-09-24)
The immunoglobulin family cell adhesion receptor L1 is induced in CRC cells at the invasive front of the tumor tissue, and confers enhanced proliferation, motility, tumorigenesis, and liver metastasis. To identify putative tumor suppressors whose expression is downregulated in L1-expressing
Milan Markićević et al.
International journal of medical sciences, 11(7), 663-673 (2014-05-21)
A role of an estrogen-regulated, autocrine motogenic factor was assumed to be a major biological role of trefoil factor 1 (TFF1) in breast cancer. TFF1 is regarded as a predictive factor for positive response to endocrine therapy in breast cancer
F B Fahlbusch et al.
Placenta, 34(11), 1027-1035 (2013-09-03)
Gastrokine-1 (GKN1) is a secreted auto-/paracrine protein, described to be expressed in the gastric mucosa. In gastric cancers GKN1 expression is commonly down-regulated. While current research focusses on the exploration of tumor-suppressive properties of GKN1 with regard to its potential
Sonam Pelden et al.
Oncology reports, 30(3), 1518-1526 (2013-07-10)
Trefoil factor 1 (TFF1) is a small secretory protein expressed in various types of carcinomas including breast cancer. The TFF1 gene contains an estrogen response element and its expression can be regulated by estrogen. Previous reports showed that TFF1 could
Rosario Aschero et al.
Pediatric blood & cancer, 71(1), e30717-e30717 (2023-10-10)
The risk of relapse in retinoblastoma is currently determined by the presence of high-risk histopathologic factors in the enucleated eye. However, the probability of developing metastatic disease is heterogeneous among these patients. Evaluating a biological marker to identify high-risk patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service