Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA003322

Sigma-Aldrich

Anti-T antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Brachyury protein antibody produced in rabbit, Anti-T protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

QQPGYSQWGWLLPGTSTLCPPANPHPQFGGALSLPSTHSCDRYPTLRSHRSSPYPSPYAHRNNSPTYSDNSPACLSMLQSHDNWSSLGMPAHPSMLPVSHNASPPTSSSQY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... T(6862)

Immunogen

Brachyury protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

T gene encodes a protein called Brachyury protein that belongs to the T-box family of transcription factors. It plays an essential role in the formation and differentiation of the mesoderm and the axial development during embryogenesis of all vertebrates. It binds to a specific palindromic T-site in the DNA via its N-terminus region called the T-box and regulates transcription of certain genes. Polymorphism in the gene has been associated with a risk of spina bifida. It plays an important role in notochord development and a duplication of this gene confers major susceptibility to familial chordoma. Defects in the gene cause SAVA (Sacral agenesis with vertebral anomalies) characterized by sacral agenesis, abnormal ossification of the vertebral bodies and a persistent notochordal canal.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84792

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liselotte E Jensen et al.
Human genetics, 115(6), 475-482 (2004-09-28)
The transcription factor T is essential for mesoderm formation and axial development during embryogenesis. Embryonic genotype for a single-nucleotide polymorphism in intron 7 of T ( TIVS7 T/C) has been associated with the risk of spina bifida in some but
Xiaohong R Yang et al.
Nature genetics, 41(11), 1176-1178 (2009-10-06)
Using high-resolution array-CGH, we identified unique duplications of a region on 6q27 in four multiplex families with at least three cases of chordoma, a cancer of presumed notochordal origin. The duplicated region contains only the T (brachyury) gene, which is
A V Postma et al.
Journal of medical genetics, 51(2), 90-97 (2013-11-21)
The T gene (brachyury gene) is the founding member of the T-box family of transcription factors and is vital for the formation and differentiation of the mesoderm and the axial development of all vertebrates. We report here on four patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service