Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA003190

Sigma-Aldrich

Anti-SCUBE1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Signal peptide, CUB and EGF-like domain-containing protein 1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

EVSHITAEFEIETKMEEASDTCEADCLRKRAEQSLQAAIKTLRKSIGRQQFYVQVSGTEYEVAQRPAKALEGQGACGAGQVLQDSKCVACGPGTHFGGELGQCVPCMPGTYQDMEGQLSCTPCPSSDGLGLPGARNVSECGGKC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCUBE1(80274)

General description

SCUBE1 (signal peptide, CUB domain, EGF-like 1) gene encodes an epidermal growth factor (EGF)-related protein that contains a CUB (C1s-like) domain at the C-terminus. It contains a signal peptide sequence at the N-terminus followed by nine tandemly arranged epidermal growth factor (EGF)-like repeats, a spacer region and three cysteine-rich repeat motifs.

Immunogen

Signal peptide, CUB and EGF-like domain-containing protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SCUBE1 (signal peptide, CUB domain, EGF-like 1) gene encodes a platelet-endothelial cell surface protein that is expressed during early embryogenesis. The protein plays a role during the early stages of central nervous system development. It also functions in postnatal cell-cell adhesions. It may also function in renal regeneration and repair.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73580

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jungang Zhao et al.
Journal of translational medicine, 20(1), 520-520 (2022-11-09)
The tumour microenvironment and cirrhotic liver are excellent sources of cancer-associated fibroblasts (CAFs), which participate in carcinogenesis. Thus, it is important to clarify the crosstalk between CAFs and HCC cells and the related mechanism in regulating carcinogenesis. Human hepatocellular carcinoma
Junli Zhuang et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 25(5), 1421-1428 (2010-01-01)
We have identified that a novel developmental gene and protein, SCUBE1, is expressed in endothelial cells and may play an important role in kidney regeneration. The temporal and spatial expression of SCUBE1 was determined in a mouse model of ischaemia-reperfusion
S Grimmond et al.
Genomics, 70(1), 74-81 (2000-11-23)
The epidermal growth factor (EGF) superfamily comprises a diverse group of proteins that function as secreted signaling molecules, growth factors, and components of the extracellular matrix, many with a role in vertebrate development. We have isolated a novel mammalian gene
Cheng-Fen Tu et al.
The Journal of biological chemistry, 283(18), 12478-12488 (2008-02-28)
SCUBE1 (signal peptide-CUB-EGF domain-containing protein 1) is a novel, secreted, cell surface glycoprotein expressed during early embryogenesis and found in platelet and endothelial cells. This protein is composed of an N-terminal signal peptide sequence followed by nine tandemly arranged epidermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service