Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA002692

Sigma-Aldrich

Anti-NFKBIE antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IκBε antibody produced in rabbit, Anti-I-κ-B-ε antibody produced in rabbit, Anti-IKB-ε antibody produced in rabbit, Anti-IKBE antibody produced in rabbit, Anti-NF-κ-B inhibitor ε antibody produced in rabbit, Anti-NF-κ-BIE antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PEPGRGTSHSLDLQLQNWQGLACLHIATLQKNQPLMELLLRNGADIDVQEGTSGKTALHLAVETQERGLVQFLLQAGAQVDARMLNGCTPLHLAAGRGLMGISSTLCKAGADSLLRNVEDETPQDLTEESLVLLPFDDLKI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKBIE(4794)

Looking for similar products? Visit Product Comparison Guide

General description

NFKBIE (NFKB inhibitor ε) belongs to the novel IκB (nuclear factor of κ light polypeptide gene enhancer in B-cells inhibitor) family and is expressed in particular cell types such as the spleen, lung and testis at different level. It is mainly involved in the regulation of NF (nuclear factor)-κB-dependent transcription.

Immunogen

NF-κ-B inhibitor ε recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NFKBIE (NFKB inhibitor ε) associates and inhibits nuclear factor κB (NF-κB), a DNA binding protein complex, which is essential for the regulation of viral genome expression. It interacts with various NF-κB proteins in vitro and in vivo, such as p65 (RelA), c-Rel, p50, and p52 to inhibit the DNA-binding activity. It restricts movement of rel protein to the cytoplasm which leads to the inhibition of NF-κB-directed transactivation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83075

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Whiteside et al.
The EMBO journal, 16(6), 1413-1426 (1997-03-17)
We have isolated a human cDNA which encodes a novel I kappa B family member using a yeast two-hybrid screen for proteins able to interact with the p52 subunit of the transcription factor NF-kappa B. The protein is found in
Z Li et al.
Molecular and cellular biology, 17(10), 6184-6190 (1997-10-07)
A novel member of the I kappaB family has been identified as a protein that associated with the p50 subunit of NF-kappaB in a yeast two-hybrid screen. Similar to previously known I kappaB proteins, this member, I kappaB epsilon, has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service