Skip to Content
MilliporeSigma
All Photos(9)

Documents

HPA002645

Sigma-Aldrich

Anti-PDIA3 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-58 kDa glucose-regulated protein antibody produced in rabbit, Anti-58 kDa microsomal protein antibody produced in rabbit, Anti-Disulfide isomerase ER-60 antibody produced in rabbit, Anti-ERp57 antibody produced in rabbit, Anti-ERp60 antibody produced in rabbit, Anti-Protein disulfide-isomerase A3 precursor antibody produced in rabbit, Anti-p58 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
RNAi knockdown
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

HMTEDNKDLIQGKDLLIAYYDVDYEKNAKGSNYWRNRVMMVAKKFLDAGHKLNFAVASRKTFSHELSDFGLESTAGEIPVVAIRTAKGEKFVMQEEFSRDGKALERFLQDYFDGNLKRYLKSEPIPESNDGP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDIA3(2923)

Looking for similar products? Visit Product Comparison Guide

General description

Protein disulfide isomerase family A, member 3 (PDIA3) is a liver microsomal protein with molecular mass of 58kDa. It is associated with the protein disulfide isomerase activity. Mature protein consists of two thioredoxin-like active site motifs (CGHC), a nuclear localization motif (KPKKKKK) at its C-terminus end and an endoplasmic reticulum-retention/retrieval motif (QEDL). It has been found in the lumen of the endoplasmic reticulum network, which is a major source for secreting disulphide-bonded polypeptides.

Immunogen

Protein disulfide-isomerase A3 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Along with the isomerase activity PDIA3 (protein disulfide isomerase family A member 3) is also involved in the carnitine acyltransferase or protease activities. It participates in different cellular functional activities: (1) hydroxylation of proline residues in procollagen, (2) disulphide bond oxidation, isomerization and reduction, (3) a major non-nuclear binding protein of the thyroid hormone 3,3′,5-triiodo-L-thyronine and (4) a component of the oligosaccharide transferase. It also acts as a chaperone, a subunit of prolyl 4-hydroxylase and microsomal triglyceride transfer protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86568

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Noiva
Seminars in cell & developmental biology, 10(5), 481-493 (1999-12-22)
Protein disulfide isomerase (PDI) is a protein-thiol oxidoreductase that catalyzes the oxidation, reduction and isomerization of protein disulfides. In the endoplasmic reticulum PDI catalyzes both the oxidation and isomerization of disulfides on nascent polypeptides. Under the reducing condition of the
J A Bassuk et al.
Matrix (Stuttgart, Germany), 9(3), 244-258 (1989-06-01)
Protein disulphide isomerase (E.C. 5.3.4.1) has been purified, cloned, and sequenced from a variety of vertebrate tissues. The enzyme and its isoforms have been assigned a role in four functional activities: (1) hydroxylation of proline residues in procollagen; (2) disulphide
M Bourdi et al.
Archives of biochemistry and biophysics, 323(2), 397-403 (1995-11-10)
The function of a 58-kDa liver microsomal protein (P58) is controversial. To help clarify the physiological function of this protein, particularly in humans, a full-length human liver cDNA clone was isolated, sequenced, and expressed in milligram quantities with the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service