Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA001522

Sigma-Aldrich

Anti-LUM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KSPG lumican antibody produced in rabbit, Anti-Keratan sulfate proteoglycan lumican antibody produced in rabbit, Anti-Lumican precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

DFPLSIYGQSSPNCAPECNCPESYPSAMYCDELKLKSVPMVPPGIKYLYLRNNQIDHIDEKAFENVTDLQWLILDHNLLENSKIKGRVFSKLKQLKKLHINHNNLTESVGPLPKSLEDLQLTHNKITKLGSFEGLVNLTF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LUM(4060)

General description

LUM (lumican) gene encodes a protein belonging to the small leucine-rich proteoglycan (SLRP) family. It contains keratin sulfate side chains and forms a major component of the cornea, dermal, and muscle connective tissues.

Immunogen

Lumican precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LUM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LUM (lumican) gene functions in the regulation of collagen assembly into fibrils in various connective tissues. It is essential in a highly organized collagenous matrix and corneal transparency development. It is also involved in epithelial cell migration and tissue repair.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84497

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albin Jeanne et al.
Scientific reports, 7(1), 7700-7700 (2017-08-11)
Lumican is a small leucine-rich proteoglycan (SLRP) being known as a key regulator of collagen fibrillogenesis. However, little attention has been given so far in studying its influence on tumor-associated matrix architecture. Here, we investigate the role of host lumican
S Chakravarti et al.
The Journal of cell biology, 141(5), 1277-1286 (1998-06-12)
Lumican, a prototypic leucine-rich proteoglycan with keratan sulfate side chains, is a major component of the cornea, dermal, and muscle connective tissues. Mice homozygous for a null mutation in lumican display skin laxity and fragility resembling certain types of Ehlers-Danlos
Meike de Wit et al.
Annals of surgical oncology, 20 Suppl 3, S348-S359 (2012-06-20)
Tumor stroma plays an important role in the progression and metastasis of colon cancer. The glycoproteins versican and lumican are overexpressed in colon carcinomas and are associated with the formation of tumor stroma. The aim of the present study was
Naoto Yamauchi et al.
Oncology letters, 21(6), 445-445 (2021-04-20)
Cancer-associated fibroblasts (CAFs) in the tumor microenvironment play an essential role in the tumor progression of esophageal squamous cell carcinoma (ESCC). The present study aimed to investigate the expression of CAF-related molecules, versican, periostin and lumican, in cancer stroma, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service