Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA001466

Sigma-Aldrich

Anti-PTPN6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hematopoietic cell protein-tyrosine phosphatase, Anti-PTP-1C, Anti-Protein-tyrosine phosphatase 1C, Anti-Protein-tyrosine phosphatase SHP-1, Anti-SH-PTP1, Anti-Tyrosine-protein phosphatase non-receptor type 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

HAGPIIVHCSAGIGRTGTIIVIDMLMENISTKGLDCDIDIQKTIQMVRAQRSGMVQTEAQYKFIYVAIAQFIETTKKKLEVLQSQKGQESEYGNITYPPAMKNAHAKASRTSSKHKEDVYENLHTKNKREEKVKKQRSADKEKSKGSLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPN6(5777)

General description

PTPN6 (protein tyrosine phosphatase, non-receptor type 6) is a 68kDa cytoplasmic protein that is predominantly expressed in hematopoietic cell development, proliferation and receptor-mediated mitogenic signaling pathways. The gene contains 17 exons spanning 17kb of DNA and is localized to chromosome 12.

Immunogen

Tyrosine-protein phosphatase non-receptor type 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-PTPN6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PTPN6 (protein tyrosine phosphatase, non-receptor type 6) gene encodes a protein that belongs to the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules involved in the regulation of several processes such as cell growth, differentiation, mitotic cycle, and oncogenic transformation. The SH2 domain of this protein interacts with and binds this protein to the substrates.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78885

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Sooman et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(5), 4479-4488 (2014-02-11)
The prognosis of high-grade glioma patients is poor, and the tumors are characterized by resistance to therapy. The aims of this study were to analyze the prognostic value of the expression of the protein tyrosine phosphatase non-receptor type 6 (PTPN6
D Banville et al.
Genomics, 27(1), 165-173 (1995-05-01)
PTPN6, a protein tyrosine phosphatase with two Src homology-2 domains, is expressed predominantly in cells of the hematopoietic lineage. In addition, cDNAs encoding isoforms of the enzyme differing from the latter at their N-terminal have been detected in normal and
Roland Lehmann et al.
Mucosal immunology, 11(3), 627-642 (2018-01-04)
Protein secretion upon TLR, TNFR1, and IFNGR ligation in the human airways is considered to be central for the orchestration of pulmonary inflammatory and immune responses. In this study, we compared the gene expression and protein secretion profiles in response
A Beghini et al.
Human molecular genetics, 9(15), 2297-2304 (2000-09-26)
The SH2 domain-containing tyrosine phosphatase PTPN6 (SHP-1, PTP1C, HCP) is a 68 kDa cytoplasmic protein primarily expressed in hematopoietic cell development, proliferation and receptor-mediated mitogenic signaling pathways. By means of direct dephosphorylation, it down-regulates a broad spectrum of growth-promoting receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service