Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA001072

Sigma-Aldrich

Anti-TYMP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ECGF1, Anti-Gliostatin, Anti-PD-ECGF, Anti-Platelet-derived endothelial cell growth factor, Anti-TP, Anti-TdRPase, Anti-Thymidine phosphorylase precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EGSQGLPDPSPEPKQLPELIRMKRDGGRLSEADIRGFVAAVVNGSAQGAQIGAMLMAIRLRGMDLEETSVLTQALAQSGQQLEWPEAWRQQLVDKHSTGGVGDKVSLVLAPALAACGCKVPMISG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TYMP(1890)

General description

The gene TYMP (thymidine phosphorylase) is mapped to human chromosome 22q13.33. It is found to be expressed in neurons of the peripheral nervous system.

Immunogen

Thymidine phosphorylase precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-TYMP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Thymidine phosphorylase is an enzyme encoded by the TYMP gene in humans. The protein stimulates angiogenesis that may occur by an indirect mechanism through its enzymatic activity. It plays an important role in angiogenesis and extracellular matrix remodeling. It is expressed in primary tumours and may be a risk factor for lymph node (LN) and hepatic metastasis in patients with gastric adenocarcinoma. The gene strongly influences gastric cancer progression by the dual activities of angiogenesis and lymphangiogenesis. TYMP is more likely expressed by malignant B cells in higher-grade lymphomas. It is an enzyme involved in nucleotide synthesis and has been implicated in critical biological processes such as DNA replication, protection against mutations and tissue repair. Deletions in this gene have been associated with mitochondrial neurogastrointestinal encephalopathy (MNGIE).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73419

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alžběta Vondráčková et al.
European journal of human genetics : EJHG, 22(3), 431-434 (2013-07-11)
Mitochondrial disorders are caused by defects in mitochondrial or nuclear DNA. Although the existence of large deletions in mitochondrial DNA (mtDNA) is well known, deletions affecting whole genes are not commonly described in patients with mitochondrial disorders. Based on the
Gene expression profiling of tuberculous meningitis co-infected with HIV.
Sameer Kumar, G. S., et al.
Journal of Proteomics and Bioinformatics, 5, 235-244 (2012)
P A Eccleston et al.
Neuroscience letters, 192(2), 137-141 (1995-06-09)
Platelet-derived endothelial cell growth factor (PD-ECGF) is an angiogenic factor which recently has been shown to be identical to thymidine phosphorylase. We describe here, high levels of expression of PD-ECGF/thymidine phosphorylase in neurons of the peripheral nervous system (PNS) but
Xianglan Zhang et al.
Pathology, 46(4), 316-324 (2014-05-07)
As an angiogenic factor, thymidine phosphorylase (TP) expression in primary tumours has been thought to be a risk factor for lymph node (LN) and hepatic metastasis in patients with gastric adenocarcinoma. However, the molecular basis for the induction of metastasis
Ghantasala S Sameer Kumar et al.
Journal of proteomics & bioinformatics, 5(9), 235-244 (2012-09-01)
Tuberculous meningitis (TBM) is a fatal form of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service