Skip to Content
MilliporeSigma
All Photos(4)

Documents

HCLBRSTDR

Sigma-Aldrich

Breast Analyte ControlDR

human cell culture

Synonym(s):

ER/PGR/HER2 Dynamic Range Analyte Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.47

product name

Breast Analyte ControlDR, Cell line controls for immunohistochemistry and in situ hybridization

biological source

human cell culture

packaging

pkg of 1 ea cell microarray block (HCL018)
pkg of 2 slides (HCL016)
pkg of 5 slides (HCL017)

manufacturer/tradename

Histocyte

storage temp.

2-8°C

Related Categories

General description

The multi-purpose Breast Analyte ControlDR has five cell line cores which demonstrate a dynamic range of expression for ER, PR, and Her-2.
HistoCyte Laboratories control slides and blocks are made from cultured human cell lines to provide the required range of expression of a specific biomarker. The cell lines are processed using proprietary methods that allow for the creation of a high density embedded cell block with cores that retain their cellular morphology. Available as pre-cut unstained slides or as cell microarray blocks for sectioning.

Application

Ideal for use as a same slide control for immunohistochemistry (IHC) and in situ hybridization (ISH) for laboratories that need a general-use breast tissue control.
The Breast Analyte ControlDR histology slides have five cell line cores providing a dynamic range of expression of ER, PGR, and Her-2 proteins. Analyte controls are placed at one end of slide, allowing sample application on same slide for valid comparison.
The Breast Analyte ControlDR cell microarray block contains five cell line cores that are 2 mm in diameter and between 3.0-3.5 mm in length. The cell microarray block may yield over 300 sections depending on thickness.

Preparation Note

Cell microarray blocks are fixed in 10% neutral buffered formalin and embedded in paraffin wax. Pre-cut unstained slides are prepared from 4 μm thick sections, mounted on positively charged slides, and dried at 37 °C overnight.

Other Notes

For certificates of analysis, contact info@histocyte.com.
Breast Analyte ControlDR brochure is available from HistoCyte Laboratories.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service