Skip to Content
MilliporeSigma
All Photos(1)

Documents

H9649

Sigma-Aldrich

Hyaluronic acid disaccharide ΔDiHA sodium salt

≥85% (HPLC)

Synonym(s):

α-ΔUA-(1→3)-GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H20NO11Na
CAS Number:
Molecular Weight:
401.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

(Rooster)

Assay

≥85% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC(C=O)C(OC1OC(=CC(O)C1O)C(O)=O)C(O)C(O)CO

InChI

1S/C14H21NO11.Na.H/c1-5(18)15-6(3-16)12(10(21)8(20)4-17)26-14-11(22)7(19)2-9(25-14)13(23)24;;/h2-3,6-8,10-12,14,17,19-22H,4H2,1H3,(H,15,18)(H,23,24);;

InChI key

SDRQEYWJTBWGNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcNAc = N-acetyl-D-glucosamine
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Preparation Note

Product of digestion of hyaluronic acid by chondroitinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service