Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

H9536

Sigma-Aldrich

Anti-Histone Deacetylase 4 (HDAC4) (DM-15) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HDAC4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~140 kDa

species reactivity

human, mouse, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 10-20 μg using RIPA extract of HeLa nuclei
indirect immunofluorescence: 1:250 using HEK 293T cells expressing recombinant mouse HDAC4
microarray: suitable
western blot: 1:1,000 using whole extracts of mouse NIH3T3 cells
western blot: 1:500 using whole extracts of rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC4(9759)
mouse ... Hdac4(208727)
rat ... Hdac4(363287)

General description

Histone deacetylases (HDACs) are competing enzymes, belonging to histone deacetylase family. There are two classes of HDACs with six to seven different types of HDACs proteins. HDAC1,HDAC2 and HDAC3 belong to Class I HDACs and HDAC4, HDAC6, and HDAC7 belong to Class II HDACs. Class I HDACs consists of a single deacetylase domain at the N-termini and diversified C-terminal regions, while Class II contains a deacetylase domain at C-terminal position.
Mammalian histone deacetylases (HDACs) are classified into three classes according to sequence homology. HDAC1, HDAC2, HDAC3 and HDAC8 belong to class I HDACs and HDAC4, HDAC6, and HDAC7 belong to class II HDACs. Class III consists of the yeast sirtuins (sir2-like protein). Class I HDACs are ubiquitously expressed. Whereas, most of the class II HDACs show tissue-specific expression. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.

Specificity

Anti-Histone Deacetylase 4 (HDAC4), recognizes human, mouse and rat HDAC4.

Immunogen

Synthetic peptide corresponding to amino acid residues of human HDAC4 with C-terminal added lysine conjugated to KLH.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below:
  • Western Blotting
  • Immnofluorescence
  • Immunoprecipitation

Biochem/physiol Actions

Histone deacetylase 4 (HDAC4) contains import and export nuclear signal that aids in nucleocytoplasmic shuttling. It has physical and functional interactions with transcription factor myocyte enhancer factor-2 (Mef2) protein and 14-3-3. This interaction disturbs nucleocytoplasmic shuttling and then guides HDAC4 to the cytoplasm and the nucleus, respectively. In addition, HDAC4 with HDAC5 and HDAC7 represses transcription by binding to the nuclear receptor co-repressors silencing mediator of retinoic acid and thyroid hormone receptors (SMRT) and nuclear receptor co-repressor (NCOR).
Studies show that HDAC4 contains a composed import and export nuclear signal that aids in nucleocytoplasmic shuttling. It has physical and functional interactions with transcription factor MEF2C and 14-3-3. This interaction results in the shuttling and it guides HDAC4 to the cytoplasm and the nucleus.The C-terminal domain of HDAC4 shows innate histone deacetylase activity. It is seen that MEF2C combines with HDAC4 at the N-terminal domain and downmodulate c-jun promoter activity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For prolonged storage, freeze in working aliquots at -20°C. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hdac4 interactions in Huntington's Disease viewed through the prism of multiomics
<BIG>Federspiel JD, et al.</BIG>
Molecular and Cellular Proteomics (2019)
Michal Mielcarek et al.
PloS one, 6(11), e27746-e27746 (2011-12-06)
Huntington's disease (HD) is a progressive neurological disorder for which there are no disease-modifying treatments. Transcriptional dysregulation is a major molecular feature of HD, which significantly contributes to disease progression. Therefore, the development of histone deacetylase (HDAC) inhibitors as therapeutics
Joel D Federspiel et al.
Molecular & cellular proteomics : MCP, 18(8 suppl 1), S92-S113 (2019-05-02)
Huntington's disease (HD) is a monogenic disorder, driven by the expansion of a trinucleotide (CAG) repeat within the huntingtin (Htt) gene and culminating in neuronal degeneration in the brain, predominantly in the striatum and cortex. Histone deacetylase 4 (Hdac4) was
Class II and IV HDACs function as inhibitors of osteoclast differentiation
Blixt NC, et al.
PLoS ONE, 12(9), e0185441-e0185441 (2017)
A H Wang et al.
Molecular and cellular biology, 19(11), 7816-7827 (1999-10-19)
Histone acetylation plays an important role in regulating chromatin structure and thus gene expression. Here we describe the functional characterization of HDAC4, a human histone deacetylase whose C-terminal part displays significant sequence similarity to the deacetylase domain of yeast HDA1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service