Skip to Content
MilliporeSigma

H7016

Hirudin

from leeches, ≥1,500 ATU/mg protein, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-279-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hirudin from leeches, lyophilized powder, ≥1,500 ATU/mg protein (ATU = antithrombin units)

biological source

animal (Hirudo verbana)

Quality Segment

form

lyophilized powder

specific activity

≥1,500 ATU/mg protein (ATU = antithrombin units)

mol wt

7 kDa

solubility

pyridine: soluble, water: soluble

storage temp.

2-8°C

General description

Hirudin is a 7 kDa acidic protein containing 65 amino acid residues that has an isoelectric point of 3.5-4.0. It is not glycosylated and lacks tryptophan, arginine and methionine residues. It is chemically stable between pH 2-12 and at temperatures up to 80°C.

Biochem/physiol Actions

The anticoagulant, hirudin, is the most potent natural inhibitor of both soluble and clot-bound thrombin. It binds thrombin with high affinity, covering more than 20% of its surface area and occluding both the active site and exosite I (fibrinogen and PAR recognition site). Hirudin blocks thrombus growth and platelet activation. It is not metabolized in the bloodstream of humans and is eliminated unchanged via kidney filtration.
The anticoagulant, hirudin, is the most potent natural inhibitor of both soluble and clot-bound thrombin. It forms a high-affinity 1:1 complex with thrombin, occluding both the proteolytic site and exosite I (fibrinogen and PAR recognition site). Hirudin blocks thrombus growth and platelet activation, and has been suggested as the clinically preferred anticoagulant (over heparin and citrate) for its specific mode of action and absence of side effects. It is not metabolized in the bloodstream of humans and is eliminated unchanged via kidney filtration.

Analysis Note

Protein determined by Lowry.

Other Notes

One unit (ATU) will neutralize one NIH unit of thrombin at 37 °C, based on direct comparison to an NIH thrombin reference standard.


Still not finding the right product?

Explore all of our products under Hirudin from leeches


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library