Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5914

Sigma-Aldrich

Nα-Methylhistamine dihydrochloride

solid

Synonym(s):

N-Methyl-1H-imidazole-4-ethanamine dihydrochloride, NAMH dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3 · 2HCl
CAS Number:
Molecular Weight:
198.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].CNCCc1c[nH]cn1

InChI

1S/C6H11N3.2ClH/c1-7-3-2-6-4-8-5-9-6;;/h4-5,7H,2-3H2,1H3,(H,8,9);2*1H

InChI key

AYUQICXJAMPXPF-UHFFFAOYSA-N

Biochem/physiol Actions

Nα-Methylhistamine dihydrochloride helps in removing or decreasing the headache in migraine prophylaxis. It is highly effective than histamine in inducing cyclic adenosine 3′,5′-monophosphate (cAMP) synthesis.
Production of N-alpha-methyl-histamine (NAMH), a histamine H(3) receptor (H3R) agonist, is promoted in Helicobacter pylori infected human gastric mucosa. NAMH acts directly on histamine H(2) receptors (H2Rs) in animals to stimulate acid secretion and to be a H2R agonist. NAMH dose dependently stimulated cAMP productions in CHO-H2R cells. This production was inhibited by famotidine but not by thioperamide. Control CHO cells were unresponsive to either histamine or NAMH. In addition, the effect of NAMH, in terms of cAMP production in CHO-H2R cells, was more potent than that of histamine-that is, with a lower EC50 concentration and higher maximal cAMP production. Both NAMH and histamine, but not R-alpha-methyl-histamine, effectively inhibited [(3)H] tiotidine binding to CHO-H2R cells. These results confirm that NAMH, which is produced in the gastric mucosa by H pylori, is a potent H2R agonist as well as a H3R agonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Quality

The product is pure based on elemental analysis results, NMR and MS spectra.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N alpha methyl histamine versus propranolol in migraine prophylaxis
Millan GRO, et al.
The Canadian Journal of Neurological Sciences, 41(2), 233-238 (2014)
Noriko Hino et al.
The Journal of pharmacology and experimental therapeutics, 375(2), 276-285 (2020-08-31)
Histamine H3 receptor antagonists/inverse agonists are known to enhance the activity of histaminergic neurons in the brain, thereby promoting arousal and cognition. Here, we report the in vitro and in vivo pharmacological profiles for a newly synthesized histamine H3 receptor
Effects of N-alpha-methyl-histamine on human H2 receptors expressed in CHO cells
Saitoh T, et al.
Gut, 50(6), 786-789 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service