Skip to Content
MilliporeSigma

H4397

TiterMax® Classic Adjuvant

liquid

Synonym(s):

Adjuvant System, TiterMax Adjuvant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

availability

not available in Japan

storage temp.

2-8°C

General description

The effectiveness of an oil emulsion vaccine greatly depends on the form and stability of the emulsion. TiterMax Classic is a water-in-oil formulation with up to 90% water and a powerful adjuvant used in many protocols. TiterMax Classic Adjuvant contains three essential ingredients: a block copolymer, CRL89-41, squalene (a metabolizable oil) and a unique microparticulate stabilizer. The emulsion is less viscous and suitable for injection through small needles.

Application

TiterMax® Classic Adjuvant has been used to boost the production of antigen-specific polyclonal IgG in rabbits.
For mice, four doses of 10 microl each into subcutaneous or muscular sites is recommended. TiterMax® was used in mice followed by isolation of CD4 T cells for epitope mapping and to test vaccines against Neisseria species.

Biochem/physiol Actions

TiterMax Classic has been especially formulated with squalene to produce stable water-in-oil emulsions with up to 90% water. It is considerably easier to emulsify TiterMax Classic than Freund′s adjuvant. The resulting emulsion is less viscous, making it easier to inject through small needles.

Legal Information

TiterMax is a registered trademark of TiterMax USA, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library