form
solution
Quality Segment
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
concentration
4 g/L
technique(s)
microbe id | staining: suitable
pH
2.5-3.5
application(s)
hematology
histology
storage temp.
room temp
Application
Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.
Other Notes
4 g/L certified hematoxylin
Still not finding the right product?
Explore all of our products under Hematoxylin Solution, Gill No. 2
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
