Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29401320

Sigma-Aldrich

StrepTrap XT prepacked chromatography column

pack of 5 × 1 mL, prepacked chromatography column

Synonym(s):

StrepTrap High Performance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106501
NACRES:
SB.53

grade

for chromatography

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 29401320

Looking for similar products? Visit Product Comparison Guide

General description

A chromatography resin for the purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.

Features and Benefits

StrepTrap XT is a ready to use column prepacked with Strep-Tactin XT Sepharose providing the benefits:
  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins.
  • Fast purifications with simple elution.
  • Physiological conditions and mild elution preserve target protein activity.
  • Fast and easy regeneration.
  • Compatible with a wide range of detergents, denaturants, and other additives.

Legal Information

StrepTrap is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service