Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9924-73

HiScreen Capto Butyl

Cytiva 28-9924-73, pack of 1 column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

butyl

packaging

pack of 1 column

manufacturer/tradename

Cytiva 28-9924-73

bed size

7.7 mm × 100 mm

column I.D.

7.7 mm

matrix

highly cross-linked agarose

avg. part. size

75 μm (d50v)

cleaning

3-14

working range

3-13

capacity

27 mg binding capacity(27 mg BSA /ml (at QB10%))

suitability

suitable for bioprocess medium

General description

Capto Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) media used in the capture and intermediate stages of protein purification.

Application

HiScreen columns are part of the process development platform available from Cytiva. The columns are prepacked with a range of BioProcess chromatography media (resins) and designed for method optimization and parameter screening. HiScreen columns have small bed volumes (4.7 ml), reducing the cost of sample and buffer consumption. The media used in HiScreen columns are also available in other prepacked formats and as bulk packs, for all scales of work from development and pilot studies to routine production.

Capto Butyl is intended for initial and intermediate step purifications requiring a chromatography medium with Low to medium hydrophobicity. Capto Butyl often works efficiently with rather Low salt concentrations. The mechanism of binding and elution onto the butyl ligand is different from that onto the phenyl ligand, giving a different selectivity. Performance during sample run and selectivity are similar to that of Butyl Sepharose 4 Fast Flow. Higher flow rates can be used with Capto Butyl during equilibration, wash and regeneration.

Features and Benefits

  • Prepacked with Capto Butyl
  • Excellent for method optimization and parameter screening due to the 10 cm bed height
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same flow velocity

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiScreen is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service