Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9042-69

illustra Plasmid Mini (50)

Cytiva 28-9042-69, pack of 1 × 50 pieces

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

packaging

pack of 1 × 50 pieces

manufacturer/tradename

Cytiva 28-9042-69

Related Categories

General description

PlasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification.

The extracted plasmid DNA resulting from performing a miniprep is called a "plasmid miniprep" and is often performed using a plasmid purification kit. Minipreps are used in the process of molecular cloning to analyze bacterial clones. When bacteria are lysed under alkaline conditions, both chromosomal DNA and protein are denatured, but the plasmid DNA remains stable. On addition of acetate-containing neutralization buffer the larger and less supercoiled genomic DNA and proteins precipitate, but the small bacterial DNA plasmids stay in the solution.

The typical plasmid DNA yield from a fresh 1.5 mL culture of E. coli containing a high copy number plasmid is approximately 9 μg. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane.

The kit contains spin columns prepacked with a novel silica membrane, suspension, lysis, and neutralization buffers containing chaotropic salts, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.

Features and Benefits

  • Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing
  • Total preparation time of less than 10 min represents a 50% reduction in protocol time
  • Increased ease of use provided by minimal changes in pipetting volume from one part of the protocol to the next
  • Fewer centrifugation steps
  • Color-coded tubes and columns simplify the protocol
  • No organic solvents such as phenol chloroform

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

8B - Non-combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page discusses methods for plasmid DNA purification and the factors impacting resulting DNA purity and yield using illustra plasmidPrep Kits from Cytiva.

This page shows possible causes and solutions for problems that may occur during plasmid DNA preparation.

Protocols

This page shows Plasmid DNA purification using illustra™ plasmidPrep Mini Spin Kit from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service