Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-4013-53

His Spintrap

Cytiva 28-4013-53

Synonym(s):

His Spintrap Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.32

manufacturer/tradename

Cytiva 28-4013-53

packaging

pkg of 50 x 100 μL

bed volume

100 μL

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-14(Ni2+-stripped medium.)

working range

3-12(Ni2+-stripped medium.)

capacity

≥40 mg binding capacity (histidine-tagged protein/ml medium)(Protein binding capacity is protein-to-protein dependent.)

General description

His SpinTrap is a prepacked, single-use spin column for purifying histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC)

* See the section for licensing information in our terms and conditions.

Purification of unclarified samples on His SpinTrap columns minimizes loss of target protein caused by manual operations such as sample precentrifugation, transfer to centrifugation tubes, and collecting supernatant. In addition, loading unclarified sample directly to His SpinTrap columns reduces sample preparation time, which minimizes degradation and oxidation of sensitive target proteins.

His SpinTrap columns are used together with a standard microcentrifuge and one purification run takes approximately 10 min. For optimal performance, use His SpinTrap together with buffers prepared using His Buffer Kit.

The columns are prepacked with Ni Sepharose High Performance, which is precharged with nickel ions for capture of histidine-tagged proteins. Leakage of nickel ions from the Ni Sepharose High Performance medium is negligible.

Features and Benefits

  • For simple, small-scale purification of histidine-tagged* proteins and rapid expression screening.
  • Ni Sepharose High Performance in prepacked spin columns.
  • Yields up to 750 μg pure protein per column.
  • Allows direct purification of both clarified and unclarified cell lysates.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.15% Kathon CG)
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva
SpinTrap is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

How to optimize purification of histidine-tagged proteins using Cytiva products.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

See All

Protocols

This page describes a small-scale purification (minipreps) of histidine-tagged recombinant proteins using His SpinTrap and His SpinTrap Kit from Cytiva.

Ni Sepharose High Performance consists of highly cross-linked 6% agarose beads (34 µm) to which a chelating group has been immobilized and subsequently charged with Ni2+ ions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service