Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-9267-01

illustra RTG RT-PCR Beads, 0.2 ML

Cytiva 27926701, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55101520
NACRES:
NA.55

form

liquid

manufacturer/tradename

Cytiva 27926701

color

colorless

General description

Ready-To-Go RT-PCR Beads are stable at room temperature and designed for performing single-tube one-step reverse transcription-PCR. Each room-temperature-stable bead contains M-MuLV Reverse Transcriptase, RNase Inhibitor, buffer, nucleotides, and Taq DNA Polymerase. The only additional reagents required are water, template RNA, and primers. The reagents are optimized for full-length cDNA synthesis to >7.5 kb and optimal sensitivity from PCR.

The Ready-To-Go Bead format significantly reduces the number of pipetting steps, thereby increasing reproducibility of the RT-PCR technique and minimizing risk of contamination and RNA degradation. Ready-To-Go RT-PCR Beads are provided in either thin walled 0.5 ml or 0.2 ml tubes compatible with most thermocyclers. The 0.2 ml tubes come assembled in a 96-well (8 × 12) plate format that allows individual strips of eight tubes to be easily removed. This flexibility allows use of either the entire 96-well plate, strips of eight or individual 0.2 ml tubes. Each package of Ready-To-Go RT-PCR Beads contains: RT-PCR beads, control reactions and pd(N)6 and oligo(dT) cDNA primers.

Features and Benefits

Pre-dispensed, freeze-dried beads that include the reagents necessary for one-step reverse transcription-PCR* with high sensitivity and reproducibility.

  • Preformulated, predispensed, single-dose, ambient-temperature-stable beads ensure greater reproducibility between reactions, minimize pipetting steps, and reduce the potential for pipetting errors and contamination.
  • Optimized as a one-tube, one-step RT-PCR* reaction for both cDNA synthesis and PCR.
  • Each lot of RT-PCR Beads is function tested for its ability to generate highly specific PCR products to ensure lot-to-lot reproducibility
  • For highly reproducible one-step reverse transcription-PCR.*

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows PCR and RT-PCR amplification troubleshooting.

Protocols

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service