Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE25-0500-75

Sigma-Aldrich

RNAspin 96 (4)

Cytiva 25-0500-75, pack of (4 preps)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.52

manufacturer/tradename

Cytiva 25-0500-75

packaging

pack of (4 preps)

capacity

100 μg binding capacity

General description

96-well format using suitable vacuum manifolds or suitable centrifuges.

Application

illustra RNAspin 96 Isolation Kit can be used to reproducibly isolate total RNA from cultured cells, tissue, bacteria or yeast. It allows for the simultaneous processing of 96 samples via either centrifugation or vacuum-based protocols that can easily fit onto existing automated systems. The isolated RNA is of a high enough quality and quantity (up to 100 μg) for multiple downstream experiments, including sensitive enzymatic applications like quantitative reverse transcription polymerase chain reaction (QRT-PCR), primer extension, and RNase protection assays. Therefore, the RNAspin 96 Kit is a flexible, high-throughput solution for total RNA isolation.

Features and Benefits

  • For fast and efficient purification in a 96-well format.
  • 96 samples can be parallel-processed with high reproducibility in under 70 min.
  • Integrated wash plate eliminates risk of contamination.
  • Flexible processing by vacuum or centrifugation.
  • DNase I treatment removes genomic DNA, leading to high-quality total RNA product.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows possible causes and solutions for problems that may occur during RNA preparation.

Protocols

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service