Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GE17-5470-15

HiScreen Capto Q ImpRes

Cytiva 17-5470-15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

packaging

pkg of 4.7 mL

manufacturer/tradename

Cytiva 17-5470-15

parameter

59 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

Related Categories

General description

HiScreen Capto Q ImpRes columns are prepacked with Capto Q ImpRes ion exchange chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

Features and Benefits

  • Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing Protein purification.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiScreen is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service