Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5123-10

benzamidin Seph 4Fast Flow H-sub

Cytiva 17-5123-10, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

p-aminobenzamidine

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-5123-10

storage condition

(0.05 M Sodium Acetate, pH 4.0 in 20% Ethanol)

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning in place

1-9

working range

2-8

storage temp.

2-8°C

Related Categories

General description

HiTrap® Benzamidine FF (high sub) is an excellent tool for removal of proteolytic activity from a protein.

Application

Benzamidine Sepharose 4 Fast FLow (high sub) is an excellent tool for removal and/or purification of trypsin and trypsin-like serine proteases, as well as, zymogens including urokinase and prekallikrein. Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates

Features and Benefits

  • Benzamidine Sepharose 4 Fast FLow (high sub) is based on highly cross-linked 4% Agarose enabling high fLow rates without losing binding capacity
  • p-aminobenzamidine (pABA) immobilized on Sepharose 4 Fast FLow matrix gives high productivity, and allows easy scale-up.
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant Proteins
  • For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to Purify or remove serine proteases with a Benzamidine Sepharose 4 Fast Flow, Benzamidine Sepharose 4 Fast Flow from Cytiva.

This page provides information about the principles and standard conditions for different purification techniques for the purification of multiprotein complexes with from Cytiva.

This page shows how to remove serine proteases, e.g. thrombin and trypsin, and zymogens from recombinant products using HiTrap Benzamidine FF and Benzamidine Sepharose 4 Fast Flow from Cytiva.

This page describes principles and standard conditions for different purification techniques of histidine-tagged proteins using Cytiva products.

See All

Protocols

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service