Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-3712-05

HisTrap Excel

Cytiva 17-3712-05, pack of 5 × 1 mL

Synonym(s):

HisTrap Excel Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-3712-05

parameter

1 ml/min - 4 ml/min flow rate
5 bar

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 6% agarose

average diameter

90 μm

cleaning

2-14

working range

2-13

capacity

≥10 mg binding capacity (histidine-tagged protein/ml sedimented medium)

Related Categories

General description

HisTrap excel columns are prepacked with Ni Sepharose excel affinity resins for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC). HisTrap excel 1 mL and 5 mL are ready-to-use IMAC columns prepacked with Ni Sepharose excel. The design of the columns in combination with the specific properties of the resin enables fast and convenient purifications. The special type of filter in the top and bottom of the columns makes it possible to load large volumes of cell-free, unclarified samples directly on the columns without causing back pressure problems. This time-saving property helps prevent degradation and loss of sensitive target proteins.

Features and Benefits

  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling
  • HisTrap excel columns allow direct purification of cell-free, unclarified samples
  • HisTrap excel columns are made of biocompatible polypropylene. The columns are delivered with a stopper on the inlet and a snap-off end on the outlet.
  • HisTrap excel columns can be operated either with a peristaltic pump or with chromatography systems such as ÄKTA systems. The columns cannot be opened or refilled.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

H2O at room temperature. Maximum flow rate will be lower when using buffers or samples with high viscosity or performing purification at low temperature. Optimal flow rate during binding is sample-dependent. During column wash and elution, a flow rate of 1 ml/min is recommended.

Legal Information

HisTrap is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

See All

Protocols

This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using HisTrap Excel from Cytiva.

This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service